Skip to Content
Merck

TMO565-0010

Nalgene® vacuum filtration system

filter capacity 150 mL, polyethersulfone, pore size 0.1 μm, case of 12 ea

Synonym(s):

bottletop filtration system, sterile filtration, Nalgene 565-0010, bottle top strainer, disposable filter unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41121800
Pore size:
0.1 μm pore size
Filter capacity:
150 mL
Packaging:
case of 12 ea
Material:
polyethersulfone , polyethersulfone membrane
Manufacturer/tradename:
Nalgene 565-0010
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyethersulfone , polyethersulfone membrane

packaging

case of 12 ea

manufacturer/tradename

Nalgene 565-0010

filter capacity

150 mL

pore size

0.1 μm pore size

General description

Increase filtration efficiency by using the Nalgene® Rapid-Flow disposable filter units featuring the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination when filtering cell culture media, serum, additives and buffers. With a graduated upper chamber and Polyethersulfone (PES) membrane this filter unit offer higher aqueous flow rates; lower extractables (no wetting agent); lower protein binding; and greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.

Application

Nalgene™ vacuum filtration system is suitable for the filtration of serum, cell culture medium, tissue culture, buffers and additives.

Features and Benefits

  • Colour coded according to membrane material
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free
  • Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit
  • Graduated Upper chamber

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® vacuum filtration system




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
TMO565-001004061841585718