TMO312104-0008
Nalgene® wide-mouth bottle
high-density polyethylene, capacity 250 mL, natural, case of 250 ea
Synonym(s):
Nalgene bottle, plastic bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
high-density polyethylene
packaging
case of 250 ea
manufacturer/tradename
Nalgene 312104-0008
capacity
250 mL
color
natural
Looking for similar products? Visit Product Comparison Guide
General description
Confidently ship and store these Thermo Scientific Nalgene Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service