TMO312002-9025
Nalgene® diagnostic bottle
natural high-density polyethylene, volume 8 mL, case of 2000 ea
Synonym(s):
PPCO bottles, small volume bottle with closure
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
natural high-density polyethylene
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 312002-9025
capacity
8 mL
volume
8 mL
Related Categories
General description
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)