Skip to Content
Merck
All Photos(3)

Documents

T9753

Sigma-Aldrich

D-Tryptophan

≥98.0% (HPLC)

Synonym(s):

(R)-2-Amino-3-(3-indolyl)propionic acid, D-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86198
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.32

product name

D-Tryptophan, ≥98.0% (HPLC)

biological source

synthetic (organic)

Assay

≥98.0% (HPLC)

form

powder

technique(s)

cell culture | embryo: suitable
cell culture | mammalian: suitable

color

white to off-white

mp

282-285 °C (dec.) (lit.)

SMILES string

N[C@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m1/s1

InChI key

QIVBCDIJIAJPQS-SECBINFHSA-N

Gene Information

human ... GRINA(2907)
mouse ... GRINA(66168)
rat ... GRINA(266668)

Looking for similar products? Visit Product Comparison Guide

General description

Tryptophan is one of the essential and standard amino acids present in the body. It is used by the cells to produce proteins.

Application

D-Tryptophan has been used to study its efficacy on dispersing biofilm in dental unit waterlines (DUWLs) in vitro.
D-Tryptophan can be used for Human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture.The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.

Biochem/physiol Actions

Low level of D-tryptophan is capable of preventing the growth of an L-tryptophan-requiring mutant of the E. coli.

Other Notes

Unnatural isomer of tryptophan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of D-Amino Acids as Biofilm Dispersing Agent in Dental Unit Waterlines
Ampornaramveth RS, et al.
International journal of dentistry, 2018(4954) (2018)
C F Morris et al.
Plant physiology, 88(2), 435-440 (1988-10-01)
An endogenous germination inhibitor(s) in wheat (Triticum aestivum L.) grain has been implicated in seed dormancy and germination, but its identity and mode of action have not been elucidated. We isolated and identified an endogenous germination inhibitor in white wheat
Fangjie Zheng et al.
Electrophoresis, 40(16-17), 2218-2226 (2019-04-27)
Establishing a simple and accurate method for Hg2+ detection is of great importance for the environment and human health. In this work, platinum nanoparticles (Pt NPs) with different capped agents and morphologies were synthesized. It was found that Pt NPs
Yu Sakurai et al.
Molecular pharmaceutics, 17(4), 1397-1404 (2020-02-25)
Peptide modification is a popular strategy for developing an active targeting lipid nanoparticle (LNP). In modifying the surface of an LNP with a peptide, the sequence and structure of the peptide strongly affects the formation of the LNP. Specifically, a
Action of D-Tryptophan on E. coli
Hatanaka M
Nature, 204(4954), 202-203 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service