Skip to Content
Merck
All Photos(4)

Key Documents

T9299

Sigma-Aldrich

β-(1,2,4-Triazol-3-yl)-DL-alanine

≥98% (TLC)

Synonym(s):

(±)-2-Amino-3-(1,2,4-triazol-3-yl)propionic acid, DL-1,2,4-Triazole-3-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4O2
CAS Number:
Molecular Weight:
156.14
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

β-(1,2,4-Triazol-3-yl)-DL-alanine,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

NC(Cc1nc[nH]n1)C(O)=O

InChI

1S/C5H8N4O2/c6-3(5(10)11)1-4-7-2-8-9-4/h2-3H,1,6H2,(H,10,11)(H,7,8,9)

InChI key

CAPORZWUTKSILW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Inhibitory histidine analog.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Beiboer et al.
Protein engineering, 9(4), 345-352 (1996-04-01)
The effect of the substitution of the active site histidine 48 by the unnatural 1,2,4-triazole-3-alanine (TAA) amino acid analogue in porcine pancreas phospholipase A2 (PLA2) was studied. TAA was introduced biosynthetically using a his-auxotrophic Escherichia coli strain. To study solely
Yutaka Ikeda et al.
Protein engineering, 16(9), 699-706 (2003-10-16)
Proteins containing unnatural amino acids have immense potential in biotechnology and medicine. We prepared several histidine analogues including a novel histidine analogue, beta-(1,2,3-triazol-4-yl)-DL-alanine. These histidine analogues were assayed for translational activity in histidine-auxotrophic Escherichia coli strain UTH780. We observed that
Younas Aouine et al.
Molecules (Basel, Switzerland), 16(4), 3380-3390 (2011-04-23)
A simple synthetic approach to racemic N-tert-butyloxycarbonyl-2-methyl-3-(1H-1,2,4-triazol-1-yl)alanine (5) in four steps and 68% overall yield starting from oxazoline derivative 1 is reported. This synthesis involves the alkylation of 1H-1,2,4-triazole with an O-tosyloxazoline derivative, followed by an oxazoline ring-opening reaction and
P Soumillion et al.
Protein engineering, 11(3), 213-217 (1998-06-05)
The only histidine residue in the H31N-H137N double mutant of phage lambda lysozyme (lambdaL), at position 48, was biosynthetically replaced by the analogue 1,2,4-triazole-3-alanine (Taz), the basicity of which is 3 pKa units lower. A histidine-auxotrophic strain was grown to
D E Levin et al.
Environmental mutagenesis, 8(1), 9-28 (1986-01-01)
The standard Salmonella tester strains used to detect base substitution mutations carry the hisG428 ochre mutation (TA102 and TA104) and the hisG46 missense mutation (TA100). These mutations can be reverted by base changes at their mutant his loci or at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service