Skip to Content
Merck
All Photos(5)

Key Documents

T8578

Sigma-Aldrich

Anti-β-Tubulin III Antibody

mouse monoclonal, 2G10

Synonym(s):

Anti-β-Tubulin Class III, Anti-Neuron-specifc class III β-tubulin, Anti-TUBB4, Anti-Tubb3, Anti-Tubulin β-4, Anti-Tubulin β-III, Anti-Tubulin, β−3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

2G10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.25-0.5 μg/mL using PC12 cell extract

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)
mouse ... Tubb3(22152)
rat ... Tubb3(246118)

General description

Tubulin β 3 class III (TUBB3) also known as β-Tubulin III, is encoded by the gene mapped to human chromosome 16q24.3. TUBB3 protein expression is restricted to neurons.

Application

Monoclonal Anti-β-Tubulin III (neuronal) antibody produced in mouse has been used in following studies:
  • Immunofluorescence.
  • Immunocytochemistry.

Biochem/physiol Actions

Tubulin β 3 class III (TUBB3) plays a vital role in nervous system development and axon guidance. Mutation of the gene encoding protein leads to the ocular motility disorder, congenital fibrosis of the extraocular muscle type 3 (CFEOM3), and various neurological syndromes. Altered expression of the protein affects microtubule dynamics and microtubule-kinesin interactions. Increased expression of TUBB3 is observed in tumor tissues. Decreased expression of TUBB3 can be considered as an important marker for poor prognosis of cutaneous malignant melanoma. TUBB3 is implicated in the suppression of invasive growth of tumor tissue;therefore, it is considered to be a potential target for development of antitumor drugs.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hua Sun et al.
Molecular neurodegeneration, 10, 36-36 (2015-08-12)
Previous studies have demonstrated that the trafficking defects of Nav1.1/Nav1.2 are involved in the dementia pathophysiology. However, the detailed mechanisms are not fully understood. Moreover, whether the impaired miRNAs regulation linked to dementia is a key player in sodium channel
Zhiqiang Zheng et al.
PloS one, 5(9), e12555-e12555 (2010-09-15)
Embryonic stem (ES) cells hold considerable promise as a source of cells with therapeutic potential, including cells that can be used for drug screening and in cell replacement therapies. Differentiation of ES cells into the somatic lineages is a regulated
Demethylation of specific Wnt/?-catenin pathway genes and its upregulation in rat brain induced by prenatal valproate exposure.
Wang Z
Anatomical Record (Hoboken, N.J. : 2007), 293(11), 1947-1953 (2010)
A Odawara et al.
Scientific reports, 6, 26181-26181 (2016-05-18)
The functional network of human induced pluripotent stem cell (hiPSC)-derived neurons is a potentially powerful in vitro model for evaluating disease mechanisms and drug responses. However, the culture time required for the full functional maturation of individual neurons and networks
Juli R Bagó et al.
Nature communications, 7, 10593-10593 (2016-02-03)
Transdifferentiation (TD) is a recent advancement in somatic cell reprogramming. The direct conversion of TD eliminates the pluripotent intermediate state to create cells that are ideal for personalized cell therapy. Here we provide evidence that TD-derived induced neural stem cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service