Skip to Content
Merck

Skip To

T5912

Tris Buffered Saline

10X solution

Synonym(s):

TBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tris Buffered Saline, 10 ×, solution

sterility

0.2 μm filtered

Quality Segment

form

solution

concentration

10 ×

color

colorless

pH

7.2—7.6

storage temp.

2-8°C

Application

Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash microtiter plates for western blot experiments and screening methods.[1]
Suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.

Other Notes

Washing buffer for alkaline phosphatase or peroxidase conjugates in Western blotting. Working solution will contain 20 mM Tris, pH approx. 7.4, and 0.9% NaCl

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6664T9039T6789
concentration

10 ×

concentration

-

concentration

-

concentration

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solution

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C

pH

7.2—7.6

pH

8.0

pH

-

pH

7.80—8.20

sterility

0.2 μm filtered

sterility

-

sterility

-

sterility

-


Still not finding the right product?

Explore all of our products under Tris Buffered Saline


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. Which is the buffer composition? Thank you!

    1 answer
    1. The composition of this product consists of 20mM Tris and 0.9% NaCl at the 1X concentration.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. I am interested in Tris Buffered Saline solution 10x (T5912) - I know this is sterile filtered but the specification sheet does not state "Sterile" or "pass" for sterility test. Do you provide a sterility test result on this product?

    1 answer
    1. While this material is sterile-filtered as a part of the production process, sterility testing is not performed on the finished product. This product is tested for Appearance, pH, Conductivity, and Dot Blot suitability. Please see the link below to review the product specifications:
      https://www.sigmaaldrich.com/specification-sheets/183/178/T5912-BULK________SIGMA____.pdf

      Helpful?

  5. What is the CAS number for T5912-1L?

    1 answer
    1. Tris Buffered Saline is not assigned a CAS (Chemical Abstract Service) number. This is a multi-component formulation. CAS numbers are assigned to individual compounds only.

      Helpful?

Reviews

No rating value

Active Filters