Skip to Content
Merck
All Photos(1)

Key Documents

T4674

Sigma-Aldrich

Trypsin solution from porcine pancreas

sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.78

biological source

Porcine pancreas

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

solution

mol wt

23.4 kDa

technique(s)

cell culture | mammalian: suitable

impurities

Porcine parvovirus, none detected (9 CFR)

pH

7.6

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestions. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Caution

This product should be stored frozen at -20°C.

Unit Definition

One BAEE unit will produce a A253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as a substrate.

Preparation Note

This is a 25 g/L porcine trypsin solution in Hank′s Balanced Salt Solution with phenol red.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana Klun et al.
Parasites & vectors, 10(1), 167-167 (2017-04-06)
Consumption of undercooked or insufficiently cured meat is a major risk factor for human infection with Toxoplasma gondii. Although horsemeat is typically consumed rare or undercooked, information on the risk of T. gondii from infected horse meat to humans is
Feriel Yekkour et al.
Veterinary parasitology, 239, 31-36 (2017-05-13)
Toxoplasmosis is a parasitic disease with worldwide distribution and a major public health problem. In Algeria, no data are currently available about genotypes of Toxoplasma gondii isolated from animals or humans. The present study assesses for the first time the
Mian Zhang et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 34(4), 726-738 (2018-11-30)
Traumatic joint injuries produce osteoarthritic cartilage manifesting accelerated chondrocyte terminal differentiation and matrix degradation via unknown cellular and molecular mechanisms. Here we report the ability of biomechanical stress to increase expression of the calcium-sensing receptor (CaSR), a pivotal driver of
Benoît X E Desbiolles et al.
Microsystems & nanoengineering, 6, 67-67 (2021-09-28)
Volcano-shaped microelectrodes (nanovolcanoes) functionalized with nanopatterned self-assembled monolayers have recently been demonstrated to report cardiomyocyte action potentials after gaining spontaneous intracellular access. These nanovolcanoes exhibit recording characteristics similar to those of state-of-the-art micro-nanoelectrode arrays that use electroporation as an insertion
B X E Desbiolles et al.
Nano letters, 19(9), 6173-6181 (2019-08-20)
Micronanotechnology-based multielectrode arrays have led to remarkable progress in the field of transmembrane voltage recording of excitable cells. However, providing long-term optoporation- or electroporation-free intracellular access remains a considerable challenge. In this study, a novel type of nanopatterned volcano-shaped microelectrode

Protocols

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Trypsin is commonly used for dissociating adherent cells from surfaces. A wide variety of trypsin solutions are available to meet your specific cell line requirements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service