Skip to Content

T4444

3, 3′,5 ,5′-Tetramethylbenzidine

peroxidase substrate, chromogenic, aqueous solution

Synonym(s):

TMB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution

Quality Segment

form

aqueous solution

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

General description

This product contains 3, 3′,5 ,5′-Tetramethylbenzidine (TMB) in a mildly acidic buffer. The substrate is supplied as a one component ready to use solution. Rate kinetics are approx. 40% faster than traditional TMB formulations. Not recommended for membrane or immunohistochemical applications that require a precipitating reaction product.

Application

3,3′,5,5′-Tetramethylbenzidine (TMB) is a peroxidase substrate suitable for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 655 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm.
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA has been used as a substrate for peroxidase:
  • to determine synthesized hyaluronan and HAS2 (hyaluronan-synthesizing enzyme) expression by the transfectant
  • to measure the levels of α-hemolysin toxin
  • in imaging bound IgGs



Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library