Skip to Content
Merck
All Photos(1)

Key Documents

T4143

Sigma-Aldrich

Triamterene

≥99%

Synonym(s):

2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-pteridinetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N7
CAS Number:
Molecular Weight:
253.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99%

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: soluble 1.2 mg/mL
DMSO: soluble 5.4 mg/mL
H2O: insoluble

originator

Wellspring

SMILES string

Nc1nc(N)c2nc(-c3ccccc3)c(N)nc2n1

InChI

1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)

InChI key

FNYLWPVRPXGIIP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Triamterene has been used to study its effects on urokinase receptor (uPAR) induction in podocytes under in vitro conditions2. Triamterene has also been identified as an activator compound from a LOPAC library screen using a fluorogenic α-glucosidase assay3.

Biochem/physiol Actions

Weak diuretic with potassium sparing properties; blocks Na+ reuptake in the kidneys.

Features and Benefits

This compound was developed by Wellspring. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

200 mg of triamterene is soluble in 4 ml of warm formic acid and yields a clear, yellow-green solution. Triamterene is also soluble in DMSO (5.4 mg/ml), and in 45% (w/v) aqueous 2-hydroxypropyl-β-cyclodextrin (1.2 mg/ml). However, it is insoluble in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Kato et al.
The American journal of physiology, 276(1 Pt 2), F62-F71 (1999-01-14)
Infusing urea into low-protein-fed mammals increases urine concentration within 5-10 min. To determine which urea transporter may be responsible, we measured urea transport in perfused IMCD3 segments [inner medullary collecting duct (IMCD) segments from the deepest third of the IMCD]
Ferruh Artunc et al.
Kidney & blood pressure research, 32(2), 119-127 (2009-04-30)
Serum- and glucocorticoid-inducible kinase 1 (SGK1) stimulates the epithelial sodium channel (ENaC), renal outer medullary K(+) channel 1, Na(+)/K(+)-ATPase and presumably the Na(+)-Cl(-) cotransporter (NCC). SGK1-deficient mice (sgk(-/-)) show a compensated salt-losing phenotype with secondary hyperaldosteronism. The present experiments explored
Ki-Woong Jeong et al.
Biochemistry, 50(22), 4843-4854 (2011-05-07)
Vascular endothelial growth factor (VEGF), which has neurotrophic and neuroprotective effects in addition to its major role in angiogenesis, interacts with Aβ and accumulates in the senile plaques of Alzheimer's disease (AD) patients' brains. It is known that Aβ binds
Balraj S Heran et al.
The Cochrane database of systematic reviews, 11, CD008167-CD008167 (2012-11-16)
Potassium-sparing diuretics, which block the epithelial sodium channel (ENaC), are widely prescribed for hypertension as a second-line drug in patients taking other diuretics (e.g. thiazide diuretics) and much less commonly prescribed as monotherapy. Therefore, it is essential to determine the
Indoleamine 2,3-dioxygenase-expressing mature human monocyte-derived dendritic cells expand potent autologous regulatory T cells: consideration of triamterene to treat lupus.
Richard E Kast et al.
Medical hypotheses, 74(5), 957-957 (2010-02-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service