Skip to Content
Merck
All Photos(3)

Key Documents

T3777

Sigma-Aldrich

1-Triacontanol

≥98% (capillary GC)

Synonym(s):

1-Hydroxytriacontane, Melissyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)28CH2OH
CAS Number:
Molecular Weight:
438.81
Beilstein:
1711965
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (capillary GC)

form

powder

mp

86-87 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO

InChI

1S/C30H62O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31/h31H,2-30H2,1H3

InChI key

REZQBEBOWJAQKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • The Relationships between Waxes and Storage Quality Indexes of Fruits of Three Plum Cultivars.: This study examines how 1-Triacontanol affects the storage and quality of plums. By analyzing waxes that include 1-Triacontanol, the research helps understand its impact on extending the shelf life of fruits, which is crucial for agricultural economics and food sustainability (Zhu et al., 2023).

  • Mitigation of salt stress in Indian mustard (Brassica juncea L.) by the application of triacontanol and hydrogen sulfide.: This research explores the use of 1-Triacontanol to alleviate salt stress in Indian mustard plants, offering insights into how it enhances plant growth under stress conditions. Such studies are vital for improving crop resilience to environmental stresses, thus contributing to sustainable agriculture practices (Verma et al., 2023).

  • The Effect of the Stress-Signalling Mediator Triacontanol on Biochemical and Physiological Modifications in Dracocephalum forrestii Culture.: The study delves into how 1-Triacontanol acts as a stress mediator in plant cultures, highlighting its potential to induce biochemical and physiological changes that could lead to enhanced secondary metabolite production in medicinal plants (Weremczuk-Jezyna et al., 2022).

  • Triacontanol priming as a smart strategy to attenuate lead toxicity in Brassica oleracea L.: This investigation shows the effectiveness of 1-Triacontanol in mitigating lead toxicity in plants, providing a strategy to protect crops from heavy metal contamination, thereby ensuring food safety and environmental health (Ahmed et al., 2023).


Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Akitha Devi et al.
Indian journal of experimental biology, 50(10), 729-734 (2012-12-12)
Development of a reproducible, versatile and efficient in vitro plant regeneration system is highly warranted for Indian soybean varieties for their mass multiplication in view of their commercial significance. Accordingly a protocol for direct shoot organogenesis in soybean variety JS
Shijie Du et al.
Natural product research, 23(14), 1316-1321 (2009-09-08)
A new lactone was isolated from the leaves of Xylocarpus granatum, along with three known compounds: triacontanol, beta-sitosterol and kaempferol-3-O-beta-D-glucoside. Its structure was elucidated as 3-(1-hydroxyethyl)-4,4-dimethyl-4-butyrolactone (1) by infrared, (1)H and (13)C NMR and ESI-MS data. At a concentration of
Z Zhang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 22(1), 28-29 (2003-02-11)
From the ethanol extract of Lysimachia pentapetala, six compounds have been isolated. These compounds were identified as triacontanol, beta-sitosterol, glycerol monopalmitate, succinic acid, flavonol glycosides and hitrite upon the date of mp, IR, MS and 1HNMR tests.
Krishnamurthy Ramanarayan et al.
Journal of plant physiology, 161(4), 489-492 (2004-05-07)
Triacontanol (TRIA), a long chain aliphatic alcohol (C30H61OH) reverses the effect of jasmonic acid (JA) in inducing proteinase inhibitors (PIs) in tomato leaves. Porcine pancreas trypsin and Spodoptera litura gut proteinases were inhibited in the presence of leaf proteins treated
Neena Srivastava et al.
Journal of chromatographic science, 47(10), 936-939 (2009-11-26)
A high-performance thin layer chromatographic method has been established for the identification and standardization of Stereospermum suaveolens extracts using two active biomarkers viz. triacontanol and p-coumaric acid in root samples collected from different geographical locations of India. These marker components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service