Skip to Content
Merck

T3196

Heptakis(2,3,6-tri-O-benzoyl)-β-cyclodextrin

>95%

Synonym(s):

β-Cyclodextrin heptakis(2,3,6-tri-O-benzoate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C182H154O56
Molecular Weight:
3237.14
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>95%

form

powder

storage temp.

2-8°C

Application

For capillary electrophoresis

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library