Skip to Content
Merck

Skip To

T3128

Trizma® maleate

Synonym(s):

TRIS maleate salt, Tris(hydroxymethyl)aminomethane maleate salt, Tromethamine maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2C(CH2OH)3 · C4H4O4
CAS Number:
Molecular Weight:
237.21
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
276-455-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.5% NaOH basis (anhydrous, titration)

Quality Segment

form

crystalline powder

color

colorless

solubility

water: 500 mg/mL, clear, colorless

SMILES string

NC(CO)(CO)CO.OC(=O)\C=C/C(O)=O

InChI

1S/C4H11NO3.C4H4O4/c5-4(1-6,2-7)3-8;5-3(6)1-2-4(7)8/h6-8H,1-3,5H2;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

HTMWOUBCEZXSHN-BTJKTKAUSA-N

Application

Trizma® maleate has been used:
  • in the rehydration of sputum cytospin samples for immunocytochemical analysis
  • in the incubation of spinal cords sections prior to nucleoside diphosphatase assay
  • as a component of alkaline phosphate (AP) staining for embryonic stem cells

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9332893337T1258
assay

≥99.5% NaOH basis (anhydrous, titration)

assay

≥99.5% (NT)

assay

≥99.0% (NT)

assay

≥99.0% (titration)

Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

300

form

crystalline powder

form

powder or crystals

form

crystalline powder

form

crystalline powder

solubility

water: 500 mg/mL, clear, colorless

solubility

H2O: 0.5 M at 20 °C, clear, colorless

solubility

H2O: 0.5 M at 20 °C, clear, colorless

solubility

water: 0.5 M, clear, colorless

color

colorless

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Trizma® maleate


Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Oral

target_organs

Central nervous system



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters