Skip to Content
Merck

T2944

Trizma® hydrochloride solution

BioPerformance Certified, suitable for cell culture, pH 7.5

Synonym(s):

Tris hydrochloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161700
EC Number:
234-318-8
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

BioPerformance Certified, Molecular Biology

form

solution

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected, bioburden, tested, endotoxin, tested, ≤5 ppm Heavy metals (as Pb)

pH

7.5

useful pH range

7.0-9.0

absorption

≤0.05 at 290 at 40%

General description

A series of Pre-mixed solutions of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy ± 0.1 pH units.

Application

Trizma® hydrochloride solution has been used as buffering agent in transcriptomics studies of kidney organoid cells.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Trizma® hydrochloride solution


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library