Skip to Content
Merck
All Photos(3)

Key Documents

T2909

Sigma-Aldrich

Tomatidine hydrochloride

≥85%

Synonym(s):

3β-Hydroxy-5α-tomatidane hydrochloride, 5α-Tomatidan-3β-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45NO2 · HCl
CAS Number:
Molecular Weight:
452.11
Beilstein:
3920740
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥85%

storage temp.

2-8°C

SMILES string

Cl[H].[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@@]4(C)[C@@]3([H])C[C@]5([H])O[C@]6(CC[C@H](C)CN6)[C@@H](C)[C@]45[H])[C@@]1(C)CC[C@H](O)C2

InChI

1S/C27H45NO2.ClH/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4;/h16-24,28-29H,5-15H2,1-4H3;1H/t16-,17-,18-,19-,20+,21-,22-,23-,24-,25-,26-,27+;/m0./s1

InChI key

SXXHVPYRDFJKPG-XRWUXUKGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tomatidine is a glycoalkaloid and an aglycone metabolite of tomatine present in high levels in unripe tomato.

Application

Tomatidine hydrochloride has been used:
  • to evaluate its effect on nasopharyngeal carcinoma using cell proliferation and flow cytometry assay
  • as a negative control in the non-specific inhibition on hedgehog signaling in hepatocellular carcinoma cell lines
  • as a control for cell growth assay and expression studies in pancreatic cancer cells

Biochem/physiol Actions

Potent inhibitor of Staphylococcus aureus small-colony variants typically seen as complications of cystic fibrosis, with little or no effect on normal S. aureus strains. Also shows antiinflammatory activity by indirectly inhibiting iNOS and COX-2 pathways.
Tomatidine induces the anti-oxidant nuclear factor E2-related factor 2 (Nrf2) and mitophagy, favoring the delay in aging process in C.elegans. It is an inhibitor of muscle atrophy and promotes anabolism in muscle by activating mammalian target of rapamycin complex 1 (mTORC1). It may be useful for treating skeletal muscle atrophy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliet Ochola et al.
Frontiers in plant science, 11, 649-649 (2020-06-27)
Potato (Solanum tuberosum) is a widely consumed staple food crop worldwide whose production is threatened by potato cyst nematodes (PCN). To infect a host, PCN eggs first need to be stimulated to hatch by chemical components in the host root
Masaru Nakayasu et al.
Plant & cell physiology, 61(1), 21-28 (2019-12-10)
Tomato plants (Solanum lycopersicum) contain steroidal glycoalkaloid α-tomatine, which functions as a chemical barrier to pathogens and predators. α-Tomatine accumulates in all tissues and at particularly high levels in leaves and immature green fruits. The compound is toxic and causes
Xiaoyun Liao et al.
Carcinogenesis, 30(1), 131-140 (2008-11-26)
Aberrant activation of hedgehog (HH) pathway has been implicated in the development of human malignancies. This study aimed at investigating the role of HH molecules in human ovarian carcinogenesis. The expression profiles of HH molecules were examined in ovarian tumor
Shuhong Huang et al.
Carcinogenesis, 27(7), 1334-1340 (2006-02-28)
Liver cancers, the majority of which are hepatocellular carcinomas (HCCs), rank as the fourth in cancer mortality worldwide and are the most rapidly increasing type of cancer in the United States. However, the molecular mechanisms underlying HCC development are not
Activation of the hedgehog pathway in human hepatocellular carcinomas
Huang S, et al.
Carcinogenesis, 27(7), 1334-1340 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service