Skip to Content
Merck

Skip To

T2877

3,3′,5-Triiodo-L-thyronine

synthetic (organic), ≥95% (HPLC), Thyroid hormone, powder

Synonym(s):

O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
229-999-3
MDL number:
Beilstein/REAXYS Number:
2710227
Biological source:
synthetic (organic)
Form:
powder
Assay:
≥95% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder

biological source

synthetic (organic)

Quality Segment

sterility

non-sterile

assay

≥95% (HPLC)

form

powder

optical activity

[α]25/D +23.0°, c = 2 in 1 N HCl: ethanol, 1:2

technique(s)

transfection: suitable

mp

234-238 °C (lit.)

solubility

4 M NH4OH in methanol: 5 mg/mL, clear to very slightly hazy, colorless to light yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1

InChI key

AUYYCJSJGJYCDS-LBPRGKRZSA-N

Gene Information

Application

3,3′,5-Triiodo-L-thyronine can be used for Insulin-like growth factor I assays performed by using a double antibody radioimmunoassay.[1] The product can also be used for in vitro transfection of 293T cells.[2]

Biochem/physiol Actions

3,3′,5-Triiodo-L-thyronine also referred to as O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine or T3 is the biologically active thyroid hormone. In hypothyroidism pateints, triiodothyronine enhances mood and neuropsychological function.[3] The product also positively regulates the hepatic FGF21 expression[2] and stimulates the metabolic breakdown of glucose, fats, and proteins by enhancing the levels of numerous metabolic enzymes such as hexokinase, liver glucose 6-phosphatase as well as mitochondrial enzymes for oxidative phosphorylation. Additionally, the product facilitates the normal body growth in children and is pivotal for central nervous system maturation in foetus.

Preparation Note

3,3′,5-Triiodo-L-thyronine dissolves in 4M NH4OH in MeOH at 5 mg/ml to yield a colorless to light yellow, clear to very slightly hazy solution.

Other Notes

This product has not been tested for cell culture. A sterile version is offered that is suitable for cell culture. The T5516 is a sterile preparation that is cell culture tested. Please review the product page for product T2877

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6397T5516T2376
biological source

synthetic (organic)

biological source

synthetic

biological source

synthetic (organic)

biological source

synthetic

assay

≥95% (HPLC)

assay

≥95%

assay

≥90% (HPLC)

assay

≥98% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

technique(s)

transfection: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

target_organs

Thyroid,Cardio-vascular system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Can you freeze stock solutions diluted to 0.5mg/ml in 4M ammonia at minus 20?

    1 answer
    1. Stock solutions can be frozen in working aliquots. It is important to avoid repeated freeze-thaw cycles. Dilute solutions may absorb to glass surfaces.

      Helpful?

  2. Is this product suitable for cell culture ? 

    1 answer
    1. This product has not been tested for cell culture. A sterile version is offered that is suitable for cell culture. The T5516 is a sterile preparation that is cell culture tested. Please review the product page for product T2877: https://www.sigmaaldrich.com/product/sigma/t2877

      Helpful?

Reviews

No rating value

Active Filters