Skip to Content
Merck

Skip To

T2528

Terbutaline hemisulfate salt

β-adrenoceptor agonist

Synonym(s):

2-t-Butylamino-1-(3,5-dihydroxyphenyl)ethanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,3-benzenediol hemisulfate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C12H19NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
274.32
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
245-386-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Terbutaline hemisulfate salt,

Quality Segment

solubility

H2O: soluble 100 mg/mL, clear to slightly hazy, colorless to yellow

originator

Novartis

SMILES string

OS(O)(=O)=O.CC(C)(C)NCC(O)c1cc(O)cc(O)c1.CC(C)(C)NCC(O)c2cc(O)cc(O)c2

InChI

1S/2C12H19NO3.H2O4S/c2*1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8;1-5(2,3)4/h2*4-6,11,13-16H,7H2,1-3H3;(H2,1,2,3,4)

InChI key

KFVSLSTULZVNPG-UHFFFAOYSA-N

Gene Information

Application

Terbutaline hemisulfate has been used as a reference standard for the determination of β agonist residues in meat4. It has also been used as a standard for developing LC-MS-MS based techniques for the analysis of veterinary drugs5.

Biochem/physiol Actions

Terbutaline is a β-adrenoceptor agonist and is known to function as a bronchodilator. Studies have reported that terbutaline is potent inhibitor of premature labor6.

Features and Benefits

To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Terbutaline hemisulfate salt dissolves in water at 100 mg/ml to yield a colorless to yellow, clear to slightly hazy solution.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I3639B8684T0050000
Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-

solubility

H2O: soluble 100 mg/mL, clear to slightly hazy, colorless to yellow

solubility

-

solubility

H2O: soluble ~33 mg/mL

solubility

-

originator

Novartis

originator

Sanofi Aventis

originator

AstraZeneca

originator

-

Gene Information

human ... ADRB1(153), ADRB2(154), ADRB3(155)

Gene Information

human ... ADRB1(153), ADRB2(154), ADRB3(155)

Gene Information

-

Gene Information

human ... ADRB2(154)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 2 - STOT SE 1

target_organs

Cardio-vascular system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters