Skip to Content
Merck
All Photos(1)

Key Documents

T2449

Sigma-Aldrich

Triglyceride Reagent

used for quantitative enzymatic determination of triglyerides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

10 mL sufficient for 50 tests

storage temp.

2-8°C

Application

The triglyceride and free glycerol kits are for the quantitative determination of glycerol, total triglycerides or free triglycerides.

Principle

The triglyceride reagent includes lipase for hydrolysis of triglycerides to glycerol.

Linkage

In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.

related product

Product No.
Description
Pricing

required but not provided

standard

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Even Fjære et al.
The Journal of nutritional biochemistry, 66, 98-109 (2019-02-19)
Low-fat diets and exercise are generally assumed to ameliorate obesity-related metabolic dysfunctions, but the importance of exercise vs. dietary changes is debated. Male C57BL/6J mice were fed a high-fat/high-sucrose (HF/HS) diet to induce obesity and then either maintained on the
Ana Guarner et al.
Developmental cell, 43(6), 689-703 (2017-12-14)
To understand the consequences of the complete elimination of E2F regulation, we profiled the proteome of Drosophila dDP mutants that lack functional E2F/DP complexes. The results uncovered changes in the larval fat body, a differentiated tissue that grows via endocycles.
Hideaki Miyoshi et al.
Journal of cellular biochemistry, 105(6), 1430-1436 (2008-11-05)
In adipocytes, lipid droplet (LD) size reflects a balance of triglyceride synthesis (lipogenesis) and hydrolysis (lipolysis). Perilipin A (Peri A) is the most abundant phosphoprotein on the surface of adipocyte LDs and has a crucial role in lipid storage and
Olga I Kubrak et al.
Frontiers in physiology, 7, 572-572 (2016-12-10)
Insects are known to respond to seasonal and adverse environmental changes by entering dormancy, also known as diapause. In some insect species, including
Haoyong Yu et al.
Journal of molecular endocrinology, 59(2), 121-128 (2017-06-02)
5-Aminolevulinic acid (5-ALA), the first compound in the porphyrin synthesis pathway, has been reported to ameliorate the diabetic state in Otsuka Long-Evans Tokushima Fatty rats by reducing fat pad weight in the retroperitoneal region. Dietary supplementation with 5-ALA has additionally

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service