T2397
Tyrode′s Salts
With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Synonym(s):
Tyrode′s buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
sterility
sterile-filtered
Quality Level
form
liquid
quality
With sodium bicarbonate
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
glucose: 1.0 g/L (Dextro
Dextro)
NaHCO3: 1.0 g/L
shipped in
ambient
General description
Tyrode′s Salts solution is designed to work with mammalian cells. This balanced salt solution can be used for diluting protein components of media of natural origin. Balanced Salt solution plays a role in providing a buffer system to maintain the medium in optimum pH range (7.2-7.6), irrigating and diluting while maintaining the intra- and extracellular osmotic balance. It facilitates the maintenance of normal cell metabolism by giving water and bulk inorganic ions. Balanced Salt solution provides the energy source for cell metabolism by supplying nutrients.
Application
Tyrode′s Salts has been used:
- to incubate sperms collected from cauda epididymis of rat, mouse, guinea pig, golden hamster and rabbit
- for the transportation of lower uterine segment (LUS) strips from women
- for the transportation of Cor.4U human-induced pluripotent stem cell (hiPSC)-derived cardiomyocytes
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wenlong Zhao et al.
Journal of cellular and molecular medicine, 22(3), 1755-1768 (2017-11-24)
Outer dense fibers (ODFs), as unique accessory structures in mammalian sperm, are considered to play a role in the protection of the sperm tail against shear forces. However, the role and relevant mechanisms of ODFs in modulating sperm motility and
Ivan Kopljar et al.
Journal of pharmacological and toxicological methods, 91, 80-86 (2018-02-09)
Calcium-based screening of hiPS-CMs is a useful preclinical safety evaluation platform with the ability to generate robust signals that facilitates high-throughput screening and data analysis. However, due to the potential inherent toxicities, it is important to understand potential effects of
Penghua Yang et al.
Nature communications, 10(1), 282-282 (2019-01-19)
Failure of neural tube closure results in severe birth defects and can be induced by high glucose levels resulting from maternal diabetes. MARCKS is required for neural tube closure, but the regulation and of its biological activity and function have
Ni-Hao Gu et al.
Reproductive biology and endocrinology : RB&E, 17(1), 66-66 (2019-08-17)
Sperm morphology mainly refers to the shape of the head, the length of the flagellar segments, including the midpiece, principal piece and end piece, and the size of the accessory structures, including axonemes, outer dense fibers (ODFs), mitochondrial sheath (MS)
Gregor Seliger et al.
European journal of obstetrics, gynecology, and reproductive biology, 225, 172-180 (2018-05-08)
The purpose of this study was to assess, if the biomechanical properties of the lower uterine segment (LUS) in women with a previous cesarean section (CS) can be determined by ultrasound (US) elastography. The first aim was to establish an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service