Skip to Content
Merck
All Photos(1)

Key Documents

T1568

Sigma-Aldrich

Thrombopoietin human

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)

Synonym(s):

MGDF, Megakaryocyte colony-stimulating factor, TPO, c-MPL ligand

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

potency

≤1.0 ng/mL ED50

mol wt

18.6 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... THPO(7066)

Looking for similar products? Visit Product Comparison Guide

Application

TPO stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo. TPO signals through the c-mpl receptor and acts as an important regulator of circulating platelets

Biochem/physiol Actions

Thrombopoeitin is a primary regulatory factor for megakaryocytopoiesis and thrombopoiesis. The mature form of TPO is a highly conserved glycoprotein, showing homology among various mammals. It is produced by liver and kidney cells. TPO stimulates growth and maturation of megakaryocytes and megakaryocytic colonies from bone marrow cultures. TPO binds and activates an 68-78 kDa glycoprotein receptor belonging to the GH family of cytokine receptors, a family that includes receptors to growth hormone (GH), erythropoietin (EPO), and prolactin (PRL). Like GH and EPO, TPO may bind to its receptor at two distinct sites, initiating receptor dimerization and activation. Analysis of mRNA indicates also the existence of a novel truncated and potentially soluble form of TPO receptor. The viral oncogene v-mlp of the myeloproliferative leukemia virus (MPLV) contains the gene sequence for the entire cytoplasmic and transmembrane domains and a portion of the extracellular domain of c-mlp (TPO receptor).

Physical form

Lyophilized from a 0.1 % TFA solution containing 250 μg bovine serum albumin

Analysis Note

The biological activity is measured in a cell proliferation assay using MO7e cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun-Mei Lu et al.
Brain, behavior, and immunity, 44, 221-234 (2014-12-03)
Neuro-inflammation plays a key role in the occurrence and development of postoperative cognitive dysfunction (POCD). Although S100A8 and Toll-like receptor 4 (TLR4) have been increasingly recognized to contribute to neuro-inflammation, little is known about the interaction between S100A8 and TLR4/MyD88
Lei Wu et al.
The Journal of experimental medicine, 211(7), 1449-1464 (2014-06-18)
Inflammatory dilated cardiomyopathy (DCMi) is a major cause of heart failure in individuals below the age of 40. We recently reported that IL-17A is required for the development of DCMi. We show a novel pathway connecting IL-17A, cardiac fibroblasts (CFs)
J Xu et al.
Cell death and differentiation, 21(8), 1229-1239 (2014-04-29)
Macrophages can be activated and regulated by high-mobility group box 1 (HMGB1), a highly conserved nuclear protein. Inflammatory functions of HMGB1 are mediated by binding to cell surface receptors, including the receptor for advanced glycation end products (RAGE), Toll-like receptor
Wenjie Guo et al.
Autophagy, 10(6), 972-985 (2014-06-01)
Nonresolving inflammation in the intestine predisposes individuals to the development of colitis-associated cancer (CAC). Inflammasomes are thought to mediate intestinal homeostasis, and their dysregulation contributes to inflammatory bowel diseases and CAC. However, few agents have been reported to reduce CAC
Ayako Nogami et al.
Oncotarget, 6(11), 9189-9205 (2015-04-01)
FLT3-ITD and FLT3-TKD are the most frequent tyrosine kinase mutations in acute myeloid leukemia (AML), with the former associated with poor prognosis. Here, we show that the PI3K inhibitor GDC-0941 or the Akt inhibitor MK-2206 induced apoptosis through the mitochondria-mediated

Articles

Read article on hematopoietic cytokines and hematopoiesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service