Skip to Content
Merck

T1283

holo-Transferrin bovine

Iron-saturated, BioReagent, suitable for cell culture

Synonym(s):

Siderophilin, Siderophilin, iron-saturated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
234-318-8
NACRES:
NA.75
MDL number:
eCl@ss:
34058011
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioReagent

assay

97-100% (agarose gel electrophoresis)

form

powder

concentration

~25 mM

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

solubility

H2O: 50 mg/mL

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

bovine ... TF(280705)

Application

Transferrin is the primary iron binding and cell delivery molecule present in serum. Serum-free cell culture systems require a delivery format for iron. Transferrin is the preferential delivery form of iron because cells process transferrin bound iron in a physiologically appropriate way through transferrin receptors on the cell surface. Bovine holo-transferrin can be used with a wide range of cells types. Bovine holo-transferrin is loaded with iron prior to use and can be added directly to iron poor or iron-free cell culture media.

Analysis Note

Purity by agarose gel electrophoresis.


Still not finding the right product?

Explore all of our products under holo-Transferrin bovine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library