Merck
T1193
Pharmed L/S tubing
size 24
Synonym(s):
Saint Gobain Tygon 06508-24, Peristaltic Pump Tubing
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
41105108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 25 feet
manufacturer/tradename
Tygon
I.D.
0.25 in. (6.4 mm)
General description
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Still not finding the right product?
Explore all of our products under Pharmed L/S tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
T1193-25FT04061834381563