Skip to Content
Merck
All Photos(4)

Key Documents

T0909

Sigma-Aldrich

Tenoxicam

NSAID

Synonym(s):

4-Hydroxy-2-methyl-N-2-pyridinyl-2H-thieno(2,3-e)-1,2-thiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O4S2
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (perchloric acid titration)

form

powder

solubility

DMF: 25 mg/mL, clear, yellow-green

SMILES string

O=C(NC1=NC=CC=C1)C2=C(O)C(SC=C3)=C3S(N2C)(=O)=O

InChI

1S/C13H11N3O4S2/c1-16-10(13(18)15-9-4-2-3-6-14-9)11(17)12-8(5-7-21-12)22(16,19)20/h2-7,17H,1H3,(H,14,15,18)

InChI key

LZNWYQJJBLGYLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tenoxicam has been used:
  • as a non-steroidal anti-inflammatory agent (NSAID) to study its effects on root gravitropism in Arabidopsis thaliana
  • as a standard in microanalysis of NSAIDs by spectrophotometry
  • to test its effect on surface potential andmembrane fluidity modification in phosphoglyceride monolayers

Biochem/physiol Actions

Tenoxicam (TX) possesses antipyretic and analgesic effects. It elicits radical scavenging activity and has the potential to treat enkylosing spondylitis, extra-articular diseases, acute gout, and rheumatic diseases. It is also effective in treating primary dysmenorrhea, postpartum uterine contraction pain, and post-operation backaches. TX is capable of inhibiting prostaglandin synthesis.
Non-steroidal antiinflammatory drug (NSAID) with comparatively low risk of renal or hepatic toxicity.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Sun Gwak et al.
International journal of pharmaceutics, 236(1-2), 57-64 (2002-03-14)
The effects of vehicles and penetration enhancers on the in vitro permeation of tenoxicam from saturated solutions through dorsal hairless mouse skin were investigated. Various types of vehicles, including ester-, alcohol-, and ether-types and their mixtures, were used as vehicles
Microanalysis of Selected NSAIDs Using the Spectrophotometric Method
Gumulka P, et al.
Engineering and Technology, 1(2), 211-221 (2020)
Katarzyna Czapla et al.
Langmuir : the ACS journal of surfaces and colloids, 26(5), 3485-3492 (2009-12-25)
Meloxicam, piroxicam, and tenoxicam belong to a highly potent oxicam group of nonsteroidal anti-inflammatory drugs. Whereas the structurally similar oxicams have different pharmacokinetics, treatment efficiency, and adverse effects, their common mechanism of action is the inhibition of a membrane enzyme
O G Nilsen
Clinical pharmacokinetics, 26(1), 16-43 (1994-01-01)
Tenoxicam is a nonsteroidal anti-inflammatory drug (NSAID) in the oxicam group. It is completely absorbed by the oral route and is about 99% protein bound in human plasma. Intake of food delays absorption without affecting bioavailability. There is no evidence
J P Gonzalez et al.
Drugs, 34(3), 289-310 (1987-09-01)
Tenoxicam is a new non-steroidal anti-inflammatory and analgesic agent of the oxicam class, and therefore closely related to piroxicam. It possesses a long half-life which enables it to be administered once daily. Clinical trials in patients with rheumatoid arthritis, osteoarthritis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service