Skip to Content
Merck

T0452

D-α-Tocotrienol

from rice, ≥75% (GC)

Synonym(s):

3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
UNSPSC Code:
12352205
NACRES:
NA.79
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rice

assay

≥75% (GC)

form

liquid

color

colorless to brown

mp

30-31 °C

storage temp.

−20°C

SMILES string

O1[C@@](CCc2c1c(c(c(c2C)O)C)C)(CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)C

InChI

1S/C29H44O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h12,14,16,30H,9-11,13,15,17-19H2,1-8H3/b21-14+,22-16+/t29-/m1/s1

InChI key

RZFHLOLGZPDCHJ-XZXLULOTSA-N

General description

Tocotrienols belong to the vitamin E family and have an unsaturated isoprenoid side chain. The various isoforms of tocotrienols differ in their methyl substitution in the chromanol head. α-Tocotrienol present in crude palm oil is regarded as a natural supplement.

Biochem/physiol Actions

α-Tocotrienol is an effective free radical scavengerand elicits protective functionality during oxidative stress in microsomes. It is a strong antioxidant, which reduces inflammation and is used in cancer treatment. α-tocotrienol supplementation may reduce cholesterol levels in hypercholesterolemia. α-tocotrienol is neuroprotective as it blocks glutamate-induced cell death in neuronal cells.
α-Tocotrienol may be used along with other isoforms to differentiate tocotrienol activities.
Tocotrienols are widely utilized for their strong antioxidant properties. The various isoforms of tocotrienols differ in their methyl substitution in the chromanol head. α-tocotrienol demonstrates neuroprotective properties by blocking glutamate-induced cell death in neuronal cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library