Skip to Content
Merck

SYNF580010

Synthware Schlenk tube with high vaccum valve

capacity 10 mL, valve L 0-4 mm, O.D. × L 22 mm × 55 mm

Synonym(s):

Synthware F580010, storage tube

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NB.43
UNSPSC Code:
41121800
Material:
clear glass , round bottom
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

clear glass , round bottom

sterility

non-sterile

feature

neck thread Internal

packaging

pack of 1 ea

manufacturer/tradename

Kemtech F580010

O.D. × L

22 mm × 55 mm

capacity

10 mL

valve L

0-4 mm

Looking for similar products? Visit Product Comparison Guide

General description

Schlenk Tube, High Vac Valve, 10mL, Valve 0-4mm, 22X55mm includes Accessories: S600004T

Application

kemetch; synthware; tube; storage tube; schlenk tube

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
SYNF580010-1EA04065270931726

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service