Skip to Content
Merck
All Photos(1)

Key Documents

SRP7783

Sigma-Aldrich

MMP-3 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

EC 3, Matrix metalloproteinase-3, Stromelysin-1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~19.5 kDa

packaging

pkg of 10 μg

technique(s)

western blot (chemiluminescent): suitable

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP3(4314)

General description

Active MMP-3 catalytic domain from human cDNA, expressed in E. coli and purified using proprietary technologies. The active MMP-3 is very useful in studying enzyme kinetics, cleave target substrates, and screen for inhibitors.
The gene JE is an early growth response gene that was first identified in mouse 3T3 cells. The gene is mapped to mouse chromosome 11. It encodes a member of the CC subfamily, the members of which contain characteristic two adjacent cysteine residues.

Application

MCP-1 has been used to induce chemotaxis in mice macrophages.
MMP-3 human has been used to develop a simple and sensitive electrogenerated chemiluminescence (electrochemiluminescence (ECL)) peptide-based bioassay for the detection of matrix metalloproteinases releasing from living cells.

Biochem/physiol Actions

Matrix metallopeptidase 3 (MMP3) has a broad substrate specificity and is capable of degrading fibronectin, laminin, collagens III, IV, IX, and X, and cartilage proteoglycans. It participates in wound repair, progression of atherosclerosis, tissue remodeling and tumor initiation. MMP3 variants can be associated with hypertrophy of interventricular septum or hypertrophic cardiomyopathy.
The gene JE, also referred to as MCP-1 (monocyte chemoattractant protein-1), encodes a secreted chemokine that functions as a chemoattractant for monocytes and may activate their anti-tumor properties. This protein may serve as a potential therapeutic anticancer agent.

Physical form

Lyophilized powder.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in sterile PBS to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Wiera et al.
Cellular and molecular life sciences : CMLS, 78(5), 2279-2298 (2020-09-23)
Learning and memory are known to depend on synaptic plasticity. Whereas the involvement of plastic changes at excitatory synapses is well established, plasticity mechanisms at inhibitory synapses only start to be discovered. Extracellular proteolysis is known to be a key
Rowan Pentz et al.
Neurobiology of disease, 148, 105150-105150 (2020-11-02)
Matrix metalloproteinase-3 (MMP-3) has been associated with risk of Alzheimer's disease (AD). In this study we introduce a novel role for MMP-3 in degrading nerve growth factor (NGF) in vivo and examine its mRNA and protein expression across the continuum
Simple and sensitive electrogenerated chemiluminescence peptide-based biosensor for detection of matrix metalloproteinase 2 released from living cells
Dang Q, et al.
Analytical and Bioanalytical Chemistry, 408(25), 7067-7075 (2016)
Jun-Jie Chen et al.
Asian Pacific journal of tropical medicine, 7(4), 297-300 (2014-02-11)
To investigate the expression and significance of MMP-3 in synovium of knee joint at different stage in osteoarthritis (OA) patients. Knee synovial tissue were collected in 90 OA patients (the OA group). Patients in the OA group was divided into
Hematopoietic-Derived Galectin-3 Causes Cellular and Systemic Insulin Resistance.
Li P
Cell, 167, 973-984 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service