Skip to Content
Merck
All Photos(1)

Key Documents

SRP6003

Sigma-Aldrich

Procalcitonin human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

CALC1, CALCA, CGRP, Calcitonin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

80 ng/mL

mol wt

12.796 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... N/A(796)

Related Categories

General description

The gene CALC1 (calcitonin 1) is mapped to human chromosome 11. CALC1 encodes for preprocalcitonin. Preprocalcitonin is cleaved by endopeptidase at the amino-terminal to produce procalcitonin.[1] Procalcitonin works as a peptide hormone and is mainly produced by the C cells of the thyroid and certain endocrine cells of the lung.[2][3] Under normal expression conditions, procalcitonin is immediately cleaved into three specific fragments, an N terminal residue, calcitonin and katacalcin.[4] Recombinant human procalcitonin produced in Escherichia coli is a single, non-glycosylated, polypeptide chain containing 116 amino acids with a molecular weight of 12,796 Daltons.

Application

Procalcitonin human has been used in in vitro vascular permeability assay.[5]

Biochem/physiol Actions

Procalcitonin is a prohormone of calcitonin.[2] Levels of unprocessed procalcitonin rise significantly after bacterial infection, trauma or shock.[6] Procalcitonin is also associated with systemic complications in humans suffering from acute pancreatitis.[7]

Physical form

Lyophilized from a 10 mM sodium phosphate buffer, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile H?O to a concentration of 0.1 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nana-Maria Wagner et al.
Anesthesia and analgesia, 124(3), 836-845 (2016-09-23)
Procalcitonin is used as a diagnostic tool for the identification and risk stratification of septic patients. Procalcitonin plasma concentrations tightly correlate with the severity of the ongoing inflammatory reaction and can rise up to 10,000-fold. Impairment of endothelial cell function
The potential role of procalcitonin and interleukin 8 in the prediction of infected necrosis in acute pancreatitis.
Rau B
Gut, 41, 832-832 (1997)
Procalcitonin as a diagnostic test for sepsis in critically ill adults and after surgery or trauma: a systematic review and meta-analysis.
Uzzan B
Critical Care Medicine, 34, 1996-1996 (2006)
Procalcitonin as an acute phase marker.
Annals of Clinical Biochemistry, 38, 483-493 (2001)
Serum procalcitonin and C-reactive protein levels as markers of bacterial infection: a systematic review and meta-analysis.
Simon L
Clinical Infectious Diseases, 39, 206-217 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service