Skip to Content
Merck
All Photos(1)

Key Documents

SRP5255

Sigma-Aldrich

HSP70, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSP70-1, HSP72, HSPA1, HSPA1A, HSPA1B. DAQB-147D11.1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~96 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HSPA1A(3303)

General description

HSP70 is a member of the heat shock protein family and is synthesized by cells of many organisms in response to stress. HSP70 is found mostly but not exclusively in the nucleus of unstressed cells. For several hours after a short heat shock, however, it is strongly concentrated in nucleoli. Nucleoli are transiently damaged by such a heat shock: their morphology changes and assembly and export of ribosomes is blocked for several hours. HSP70 helps to stabilize the morphological changes of the nucleoli.
HSP70 (heat shock 70kDa)/Hsp70A1A (heat shock 70kDa protein 1A) is also termed as HSP72. It belongs to the molecular chaperones family. This gene is located on human chromosome 6p21.

Biochem/physiol Actions

HSP70 (heat shock 70kDa)/Hsp70A1A (heat shock 70kDa protein 1A) participates in several cellular processes, such as thermotolerance, clearance of protein aggregate, survival and proliferation. Mutation in Hsp70 gene locus is linked with systemic lupus erythematosus. Overexpression of this protein is observed in valproic acid (VPA)-resistant cell lines.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engagement of Components of DNA-Break Repair Complex and NF kappaB in Hsp70A1A Transcription Upregulation by Heat Shock
Hazra J, et al.
Testing, 12(1), e0168165-e0168165 (2017)
The impact of heat shock protein 70 gene variations on clinical presentation and outcome in schizophrenic inpatients
Pae C U, et al.
Neuropsychobiology, 59(3), 135-141 (2009)
HSP72 functionally inhibits the anti-neoplastic effects of HDAC inhibitors
Fujii K, et al.
Journal of Dermatological Science, 90(1), 82-89 (2018)
Polymorphisms in the Hsp70 gene locus are genetically associated with systemic lupus erythematosus
Furnrohr BG, et al.
Annals of the Rheumatic Diseases, annrheumdis122630-annrheumdis122630 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service