Skip to Content
Merck
All Photos(1)

Documents

SRP3174

Sigma-Aldrich

TIMP-2 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

CSC-21K, Tissue inhibitor of metalloproteinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

0.5 μg/mL

mol wt

21.8 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIMP2(7077)

General description

Tissue inhibitor of metalloproteinase-2 (TIMP-2) is an extracellular inhibitor of matrix metalloproteinases (MMPs) including MMP1, 2, 3, 7, 8, 9, 10, 12, 13, 14, 15, 16 and 19. It belongs to the I35 (TIMP) family of irreversible protease inhibitors. The gene encoding this protein is localized on human chromosome 17q25. Recombinant human TIMP-2 is a 21.8kDa protein containing 194 amino acid residues.

Biochem/physiol Actions

Tissue inhibitor of metalloproteinase-2 (TIMP-2), like other protease inhibitors, functions as a key modulator of extracellular matrix degradation during tissue development and remodeling. TIMP-2 can also act through a matrix metalloproteinase (MMP)-independent mechanism inhibiting endothelial cell proliferation in vitro. It demonstrates anti-angiogenic activities in vivo. TIMP-2 is a target gene of the microRNA-miR-22.

Sequence

MCSCSPVHPQ QAFCNADVVI RAKAVSEKEV DSGNDIYGNP IKRIQYEIKQ IKMFKGPEKD IEFIYTAPSS AVCGVSLDVG GKKEYLIAGK AEGDGKMHIT LCDFIVPWDT LSTTQKKSLN HRYQMGCECK ITRCPMIPCY ISSPDECLWM DWVTEKNING HQAKFFACIK RSDGSCAWYR GAAPPKQEFL DIEDP

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Wang et al.
PloS one, 9(5), e96982-e96982 (2014-05-09)
Vasculogenic mimicry (VM) is a newly-defined tumor microcirculation pattern in highly aggressive malignant tumors. We recently reported tumor growth and VM formation of gallbladder cancers through the contribution of the ephrin type a receptor 2 (EphA2)/focal adhesion kinase (FAK)/Paxillin signaling
Genetics of Moyamoya disease.
Roder C
Journal of Human Genetics, 55(11), 711-716 (2010)
D E Gomez et al.
European journal of cell biology, 74(2), 111-122 (1997-11-14)
Four members of the tissue inhibitor of metalloproteinases (TIMP) family have been characterized so far, designated as TIMP-1, TIMP-2, TIMP-3, and TIMP-4. TIMP-1 and TIMP-2 are capable of inhibiting the activities of all known matrix metalloproteinases (MMPs) and as such
Growth-stimulatory activity of TIMP-2 is mediated through c-Src activation followed by activation of FAK, PI3-kinase/AKT, and ERK1/2 independent of MMP inhibition in lung adenocarcinoma cells.
Kim Hle
Oncotarget, 6(40), 42905-42922 (2015)
Promotion of astrocytoma cell invasion by micro RNA-22 targeting of tissue inhibitor of matrix metalloproteinase-2.
Ohnishi YI
Journal of Neurosurgery. Spine, 26(3), 396-403 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service