Skip to Content

SML4074

Trabectedin

≥98% (HPLC)

Synonym(s):

(1′R,6R,6aR,7R,13S,14S,16R)-6′,8,14-Trihydroxy-7′,9-dimethoxy-4,10,23-trimethyl-19-oxo-3′,4′,6,7,12,13,14,16-octahydrospiro(6,16-(epithiopropanooxymethano)-7,13-imino-6aH-1,3-dioxolo(7,8)isoquino(3,2-b)(3)benzazocine-20,1′(2′H)-isoquinolin)-5-yl acetate, 1′R,6R,6aR,7R,13S,14S,16R)-5-(Acetyloxy)-3′,4′,6,6a,7,13,14,16-octahydro-6′,8,14-trihydroxy-7′,9-dimethoxy-4,10,23-trimethylspiro[6,16-(epithiopropanoxymethano)-7,13-imino-12H-1,3-dioxolo[7,8]isoquino[3,2-b][3]benzazocine-20,1′(2′H)-isoquinolin]-19-one, ET-743, Ecteinascidin 743

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C39H43N3O11S
CAS Number:
Molecular Weight:
761.84
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC(C1=C2OCO1)=C(OC(C)=O)C3=C2[C@H](CO4)N([C@@H](O)[C@H]5N(C)[C@@H]6C7=C(C5)C=C(C)C(OC)=C7O)C6[C@H]3SC[C@@]8(C4=O)NCCC9=C8C=C(OC)C(O)=C9

InChI

1S/C39H43N3O11S/c1-16-9-20-10-22-37(46)42-23-13-50-38(47)39(21-12-25(48-5)24(44)11-19(21)7-8-40-39)14-54-36(30(42)29(41(22)4)26(20)31(45)32(16)49-6)28-27(23)35-34(51-15-52-35)17(2)33(28)53-18(3)43/h9,11-12,22-23,29-30,36-37,40,44-46H,7-8,10,13-15H2,1-6H3/

InChI key

PKVRCIRHQMSYJX-CXIMGACLSA-N

Biochem/physiol Actions

Anti-proliferative; a potent anticancer agent.


Trabectedin is an alkylating drug that binds guanine residues in the minor groove of DNA, causing RNA polymerase II arrest, interfering with transcription and DNA repair pathways. It is anti-proliferative, a potent anticancer agent that has been approved to treat advanced soft tissue sarcoma and is being investigated for treatment of multiple other cancers.


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Muta. 1B - Repr. 2 - STOT RE 2 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library