Skip to Content
Merck

SML3844

Iberin

≥97% (NMR)

Synonym(s):

1-Isothiocyanato-3-(methylsulfinyl)propane, 3-Methylsulfinylpropyl isothiocyanate, NSC 321801

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H9NOS2
CAS Number:
Molecular Weight:
163.26
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥97% (NMR)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (NMR)

form

liquid

color

colorless to yellow

storage temp.

-10 to -25°C

SMILES string

[S](=O)(CCCN=C=S)C

InChI key

LELAOEBVZLPXAZ-UHFFFAOYSA-N

Biochem/physiol Actions

Iberin is a natural isothiocyanates from cruciferous vegetables (cabbage, red cabbage, horseradish) that potently inhibits toll-like receptors (TLRs) signaling. Iberin covalently binds to TLR4 and TLR2, which inhibits TLR dimerization. Iberin exhibits potent antioxidant, anticancer, and antimicrobial activities. It is quorum-sensing inhibitor (QSI) of the bacterial pathogen Pseudomonas aeruginosa through downregulation of the Gac/Rsm QS network.
Inhibits toll-like receptors (TLRs); natural isothiocyanates from cruciferous vegetables


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library