Skip to Content
Merck
All Photos(2)

Key Documents

SML3778

Sigma-Aldrich

T-5224

≥98% (HPLC)

Synonym(s):

3-[5-(4-Cyclopentyloxy-2-hydroxybenzoyl)-2-[(3-oxo-1,2-benzoxazol-6-yl)methoxy]phenyl]propanoic acid, 3-{5-[4-(Cyclopentyloxy)-2-hydroxybenzoyl]-2-[(3-hydroxy-1,2-benzisoxazol-6-yl)methoxy]phenyl} propionic acid, 5-[4-(Cyclopentyloxy)-2-hydroxybenzoyl]-2-[(2,3-dihydro-3-oxo-1,2-benzisoxazol-6-yl)methoxy]benzenepropanoic acid, T 5224, T5224

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H27NO8
CAS Number:
Molecular Weight:
517.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1[o]c2c([c]1=O)ccc(c2)COc3c(cc(cc3)C(=O)c4c(cc(cc4)OC5CCCC5)O)CCC(=O)O

InChI

1S/C29H27NO8/c31-24-15-21(37-20-3-1-2-4-20)8-10-22(24)28(34)19-6-11-25(18(14-19)7-12-27(32)33)36-16-17-5-9-23-26(13-17)38-30-29(23)35/h5-6,8-11,13-15,20,31H,1-4,7,12,16H2,(H,30,35)(H,32,33)

InChI key

DALCQQSLNPLQFZ-UHFFFAOYSA-N

Biochem/physiol Actions

T-5224 is an orally active, selective AP-1 transcription complex c-Fos/c-Jun inhibitor that blocks c-Fos and c-Jun DNA-binding activity, but not that of C/EBPα, ATF-2 (bZIP domain), MyoD (bHLH domain), Sp-1 (ZF domain) and NF-κB/p65 (RHD). T-5224 inhibits PMA-induced Fos/AP-1 promoter activity (IC50 ~4 μM by NIH/3T3 reporter assay) without affectingTNFα-stimulated NF-kB activity or the cellular levels of c-Fos family protein members. T-5224 exhibits therapeutic efficacy against collagen-induced arthritis (CIA) in mice in vivo (0.3-30 mg/kg/d p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Zhao et al.
Translational oncology, 14(8), 101100-101100 (2021-05-17)
Previous studies have shown that expression of activator protein-1 (AP-1) family is significantly elevated in triple-negative breast cancer (TNBC), compared with that in other breast cancer subtypes. Here we investigated the anti-tumor effect and mechanism of T-5224, an inhibitor of
Hongjie Zhang et al.
Cardiovascular research, 119(2), 536-550 (2022-06-01)
Post-natal maturation of mammalian cardiomyocytes proceeds rapidly after birth, with most of the myocytes exiting cell cycle, becoming binucleated, and adopting oxidative phosphorylation as the primary metabolic route. The triggers and transcriptional programmes regulating cardiomyocyte maturation have not been fully
Inhibition of c-Fos expression attenuates IgE-mediated mast cell activation and allergic inflammation by counteracting an inhibitory AP1/Egr1/IL-4 axis
Journal of Translational Medicine, 19(1), 261-261 (2021)
c-Fos is a mechanosensor that regulates inflammatory responses and lung barrier dysfunction during ventilator-induced acute lung injury
BMC Pulmonary Medicine, 22(1), 9-9 (2022)
Yukihiko Aikawa et al.
Nature biotechnology, 26(7), 817-823 (2008-07-01)
To inhibit arthritis upstream of inflammatory cytokine release and matrix metalloproteinase (MMP) action, we designed de novo a small-molecule inhibitor of c-Fos/activator protein-1 (AP-1) using three-dimensional (3D) pharmacophore modeling. This model was based on the 3D structure of the basic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service