Skip to Content
Merck
All Photos(2)

Key Documents

SML3744

Sigma-Aldrich

Ac-FLTD-CMK

≥95% (HPLC)

Synonym(s):

N-acetyl-Phe-Leu-Thr-Asp-chloromethylketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37ClN4O8
CAS Number:
Molecular Weight:
569.05
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ac-FLTD-CMK is a selective gasdermin D (GSDMD) clevage inhibitor (IC50 = 46.7 nM, 329 nM, 1.49 μM, respectively, against GSDMD clevage by human caspase-1, -5, -4 in cell-free enzymatic assays) that suppresses pyroptosis downstream of both canonical and noncanonical inflammasomes, as well as reduces IL-1β release following activation of the NLRP3 inflammasome in macrophages (10 μM 4h post 200 ng/mL LPS and 0.5h before ATP or nigericin). Ac-FLTD-CMK selectively blocks GSDMD clevage by inflammatory caspases without affecting apoptotic caspases (caspase-3 IC50 >100 μM) or apoptosis.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanie Quach et al.
Mucosal immunology, 12(2), 323-339 (2018-10-27)
During invasion, Entamoeba histolytica (Eh) encounter macrophages and activate them to elicit tissue damaging pro-inflammatory responses. When Eh binds macrophages via the Gal-lectin, surface EhCP-A5 RGD sequence ligates α5β1 integrin to activate caspase-1 in a complex known as the NLRP3
Endothelial cell pyroptosis plays an important role in Kawasaki disease via HMGB1/RAGE/cathespin B signaling pathway and NLRP3 inflammasome activation
Cell Death & Disease, 10(10), 10(10)-10(10) (2019)
X X Tian et al.
International endodontic journal, 54(6), 916-925 (2020-12-31)
To explore the presence and function of NLRP6-caspase 4 inflammasome in human pulp tissue and human dental pulp cells (HDPCs). Pulp tissue was collected from freshly extracted human caries-free third molars and third molars with irreversible pulpitis. Quantitative real-time polymerase
Pengfei Wang et al.
Neuroreport, 32(3), 188-197 (2021-01-21)
Pyroptosis has been reported to contribute to the traumatic brain injury (TBI) process. Ac-FLTD-CMK is a newly synthesized pyroptosis inhibitor. However, whether Ac-FLTD-CMK inhibits pyroptosis and plays a neuroprotective role after TBI is unknown. The present study aimed to determine
Jie Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(26), 6792-6797 (2018-06-13)
The inflammasomes are signaling platforms that promote the activation of inflammatory caspases such as caspases-1, -4, -5, and -11. Recent studies identified gasdermin D (GSDMD) as an effector for pyroptosis downstream of the inflammasome signaling pathways. Cleavage of GSDMD by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service