Skip to Content
Merck
All Photos(2)

Key Documents

SML3652

Sigma-Aldrich

TASP0277308

≥98% (HPLC)

Synonym(s):

(R)-3,4-Dichloro-N-(1-(4-ethyl-5-(3-(4-methylpiperazin-1-yl)phenoxy)-4H-1,2,4-triazol-3-yl)ethyl)benzenesulfonamide, 3,4-Dichloro-N-[(1R)-1-[4-ethyl-5-[3-(4-methyl-1-piperazinyl)phenoxy]-4H-1,2,4-triazol-3-yl]ethyl]benzenesulfonamide, TASP 0277308, TASP-0277308

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28Cl2N6O3S
CAS Number:
Molecular Weight:
539.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active, highly S1P1 subtype-selective sphingosine-1-phosphate (S1P) receptor antagonist with in vitro and in vivo efficacy.
TASP0277308 is an orally active, highly S1P1 subtype-selective sphingosine-1-phosphate (S1P) receptor antagonist (IC50 = 2.7/4.2 nM against 0.1 nM S1P for binding mouse/human S1P1; IC50 >8.7 μM for S1P2-5). TASP0277308 effectively inhibits S1P1-mediated GTPγS membrane binding, cAMP response and chemotaxis (IC50 = 7.8, 3.5, 15 nM, respectively, agianst 10 nM S1P; hS1P1-expressing HEK293 or CHO) and demonstrates in vivo efficacy in various animal disease models, including collagen-induced arthritis and tumor angiogenesis in mice (10-100 mg/kg bid po), as well as neuropathic pain in rats (0.6-3 mg/kg via acute po or ip, acute 30 nmol or daily 10 nmol intrathecal injection).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine 1-Phosphate Receptor Subtype 1 as a Therapeutic Target for Brain Trauma
Journal of Neurotrauma, 35(13), 1452-1466 (2018)
Sphingosine-1-phosphate receptor 1 activation in astrocytes contributes to neuropathic pain
Proceedings of the National Academy of Sciences of the USA, 116(21), 10557-10562 (2019)
Blocking S1P interaction with S1P? receptor by a novel competitive S1P?-selective antagonist inhibits angiogenesis
Biochemical and Biophysical Research Communications, 419(4), 754-760 (2012)
Targeting the S1P/S1PR1 axis mitigates cancer-induced bone pain and neuroinflammation
Pain, 158(9), 1733-1742 (2017)
Amelioration of collagen-induced arthritis by a novel S1P1 antagonist with immunomodulatory activities
Journal of immunology (Baltimore, Md. : 1950), 188(1), 206-215 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service