Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(C13H27N.C12H27N2.C3H7N.C3H5ClO.Cl)x.xHCl
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.21
eCl@ss:
32160703
Form:
powder
Storage condition:
desiccated
form
powder
Quality Segment
mol wt
581.79 g/mol
storage condition
desiccated
color
white to beige
storage temp.
-10 to -25°C
SMILES string
C=CCNCCCCCCCCCC.C=CCNCCCCCC[N+](C)(C)C.NCC=C.ClCC1OC1.[xHCl].[Cl-]
Biochem/physiol Actions
Colesevelam hydrochloride, a polyamine polymer, is an oral, non-absorbed bile acid sequestrant. Colesevelam hydrochloride absorbs bile acids in the intestine, which increases demand for cholesterol synthesis and affectively lowers low-density lipoprotein cholesterol (LDL-C) blood levels.
Oral, non-absorbed bile acid sequestrant
Disclaimer
Hygroscopic
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.