Skip to Content
Merck

SML3452

CDDO-Im

≥98% (HPLC)

Synonym(s):

(4aR,6aS,6aR,6bR,8aS,12aS,14bS)-8a-(imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-4a,5,6,6a,7,8,9,10,12,12a-decahydropicene-2-carbonitrile, 1-(2-Cyano-3,12-dioxooleana-1,9-dien-28-oyl) imidazole, CDDO-Imidazole, CDDO-imidazolide, NSC727038

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H43N3O3
CAS Number:
Molecular Weight:
541.72
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[n]6(cncc6)C(=O)[C@]21[C@H]([C@@H]3[C@]([C@@]4(CC[C@@H]5[C@@](C4=CC3=O)(C=C(C(=O)C5(C)C)C#N)C)C)(CC2)C)CC(CC1)(C)C

InChI

1S/C34H43N3O3/c1-29(2)10-12-34(28(40)37-15-14-36-20-37)13-11-33(7)26(22(34)18-29)23(38)16-25-31(5)17-21(19-35)27(39)30(3,4)24(31)8-9-32(25,33)6/h14-17,20,22,24,26H,8-13,18H2,1-7H3/t22-,24-,26-,31-,32+,33+,34-/m0/s1

InChI key

ITFBYYCNYVFPKD-FMIDTUQUSA-N

Biochem/physiol Actions

Bifunctional CDDO analog that enhances NRF2-mediated antioxidant and anti-inflammatory activity by forming KEAP1 and GSTP covalent adducts.
CDDO-Im is a bifunctional CDDO analog that enhances NRF2-mediated antioxidant and anti-inflammatory activity by forming not only KEAP1 covalent adducts (Michael adducts via cysteines, acyl adducts via lysines and tyrosines) in the same manner as CDDO and CDDO-Me, but also by covalently transacylating arginine and serine residues in GSTP and cross-linking them to adjacent cysteine residues. CDDO-Im is typically used in the range of 1-40 nM in cultures, 2-5.5 mg/kg via i.p. and 2.5 mg/kg via i.v. in vivo (mice and rats).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library