Skip to Content
Merck

SML3448

Propidium monoazide

≥98% (HPLC), Fluorescent DNA cross-linker, powder

Synonym(s):

3-Amino-8-azido-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridium dichloride, Propidium monoazide dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H32N6·Cl2
CAS Number:
Molecular Weight:
511.49
UNSPSC Code:
12352125
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Propidium monoazide, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

red to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[N-]=[N+]=NC1=CC2=C(C=C1)C3=C(C=C(C=C3)N)[N+](CCC[N+](CC)(C)CC)=C2C4=CC=CC=C4.[Cl-].[Cl-]

InChI key

NUMTTYQOTJKZSE-UHFFFAOYSA-M

Biochem/physiol Actions

Propidium derivative that allows light-induced crosslinkage to DNA for highly selective live-dead cells analysis by PCR or fluorescence microscopy.
Propidium monoazide (PMA) is a propidium derivative with an azide group that allows crosslinkage to DNA upon light exposure. PMA is only permeable to bacteria with compromised membrane integrity, making it highly selective for sick/dying/dead cells over healthy cells. PMA is typically added to cell samples (50 μM for 1-15 min in the dark) prior to cross-linking induction (650-W halogen light source), washing and DNA extraction. Due to its higher charge, PMA (+2) is less permeant to live cells and more selective than ethidium monoazide/EMA (+1) for dead cells. The stained cells can also be viewed and analyzed by fluorescence microscopy.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library