Skip to Content
Merck
All Photos(1)

Key Documents

SML3423

Sigma-Aldrich

ARV-825

≥98% (HPLC)

Synonym(s):

(6S)-4-(4-chlorophenyl)-N-[4-[2-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]ethoxy]ethoxy]ethoxy]ethoxy]phenyl]-2,3,9-trimethyl-6H-Thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H47ClN8O9S
CAS Number:
Molecular Weight:
923.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC=C(C2=N[C@@H](CC(NC3=CC=C(C=C3)OCCOCCOCCOCCNC4=C(C5=CC=C4)C(N(C5=O)C6C(NC(CC6)=O)=O)=O)=O)C7=NN=C(C)N7C8=C2C(C)=C(C)S8)C=C1

InChI key

RWLOGRLTDKDANT-TYIYNAFKSA-N

Biochem/physiol Actions

ARV-825 is a hetero-bifunctional PROteolysis TArgeting Chimera (PROTAC) containing a BRD4 binding group (OTX-015) on one end, a linker arm, and a cereblon (CRBN) ubiquitin E3 ligase binding group (Pomalidomide) on the other end, resulting in BRD4 ubiquitylation and degradation by the proteasome. In Burkitt’s lymphoma cells, ARV-825 was found to cause prolonged BRD4 downregulation, reduce c-Myc levels, block cell proliferation, and induce apoptosis with improved results compared to BRD4 inihbitors such as JQ1 and OTX015. It showed similar results.
Hetero-bifunctional compound with a BRD4 binding group and a ubiquitin E3 ligase binding group, resulting in BRD4 ubiquitylation and degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Pia Abruzzese et al.
Journal of hematology & oncology, 9(1), 134-134 (2016-12-03)
Anti-cancer immune responses may contribute to the control of tumors after conventional chemotherapy, and different observations have indicated that chemotherapeutic agents can induce immune responses resulting in cancer cell death and immune-stimulatory side effects. Increasing experimental and clinical evidence highlight
D T Saenz et al.
Leukemia, 31(9), 1951-1961 (2017-01-04)
The PROTAC (proteolysis-targeting chimera) ARV-825 recruits bromodomain and extraterminal (BET) proteins to the E3 ubiquitin ligase cereblon, leading to degradation of BET proteins, including BRD4. Although the BET-protein inhibitor (BETi) OTX015 caused accumulation of BRD4, treatment with equimolar concentrations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service