Skip to Content
Merck
All Photos(1)

Key Documents

SML3410

Sigma-Aldrich

EMD638683

≥98% (HPLC)

Synonym(s):

EMD 638683, EMD-638683, N′-[2-(3,5-Difluorophenyl)-2-hydroxyacetyl]-2-ethyl-4-hydroxy-3-methylbenzohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18F2N2O4
CAS Number:
Molecular Weight:
364.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=C(C(C(NNC(C(C2=CC(F)=CC(F)=C2)O)=O)=O)=CC=C1O)CC

InChI

1S/C18H18F2N2O4/c1-3-13-9(2)15(23)5-4-14(13)17(25)21-22-18(26)16(24)10-6-11(19)8-12(20)7-10/h4-8,16,23-24H,3H2,1-2H3,(H,21,25)(H,22,26)

InChI key

SSNAPUUWBPZGOY-UHFFFAOYSA-N

Biochem/physiol Actions

EMD638683 is an orally active and highly selective serum/glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 = 3 μM against HeLa cellular NDRG1 phosphorylation) that significantly decreased blood pressure in fructose-treated mice but not in control saline-treated or in SGK1-knockout animals (4460 ppm in chow, ~600 mg/kg/day). EMD638683 promotes radiation-induced suicidal death of CaCo-2 colon tumor cells in vitro (50 μM) and decreases the number of colonic tumors following chemical carcinogenesis in vivo (4460 ppm in chow).
Orally active and highly selective serum/glucocorticoid-regulated kinase 1 (SGK1) inhibitor with anti-cancer and anti-hypertensive efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-fat diet-induced activation of SGK1 promotes Alzheimer's disease-associated tau pathology
Human Molecular Genetics, 30(18), 1693-1710 (2021)
Inhibition of colonic tumor growth by the selective SGK inhibitor EMD638683
Cellular Physiology and Biochemistry, 32(4), 838-848 (2013)
Serum- and glucocorticoid-inducible kinase 1 promotes insulin resistance in adipocytes via degradation of insulin receptor substrate 1
Diabetes & metabolism journal, 37(4), e3451-e3451 (2021)
An increase in alveolar fluid clearance induced by hyperinsulinemia in obese rats with LPS-induced acute lung injury
Respiratory Physiology & Neurobiology, 279279, 103470-103470 (2020)
SGK1.1 limits brain damage after status epilepticus through M current-dependent and independent mechanisms
Neurobiology of Disease, 153, 105317-105317 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service