Skip to Content
Merck
All Photos(1)

Key Documents

SML3379

Sigma-Aldrich

D159687

≥98% (HPLC)

Synonym(s):

1-(4-((3′-Chloro-6-methoxy-[1,1′-biphenyl]-3-yl)methyl)phenyl)urea, D 159687, N-[4-[(3′-Chloro-6-methoxy[1,1′-biphenyl]-3-yl)methyl]phenyl]urea, [4-(3′-Chloro-6-methoxy-biphenyl-3-ylmethyl)phenyl]urea, [4-[[3-(3-Chlorophenyl)-4-methoxyphenyl]methyl]phenyl]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19ClN2O2
CAS Number:
Molecular Weight:
366.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

Clc1cc(ccc1)c2c(ccc(c2)Cc3ccc(cc3)NC(=O)N)OC

InChI

1S/C21H19ClN2O2/c1-26-20-10-7-15(12-19(20)16-3-2-4-17(22)13-16)11-14-5-8-18(9-6-14)24-21(23)25/h2-10,12-13H,11H2,1H3,(H3,23,24,25)

InChI key

RJJLUTWHJUDZFP-UHFFFAOYSA-N

Biochem/physiol Actions

D159687 is a brain-penetrant, orally available, highly potent and selective negative allosteric modulator (NAM) against phosphodiesterase 4 (PDE4) subtype PDE4D (hPDE4D7 IC50 = 27 nM; PDE4A1/B1/C1 IC50 = 2.50/1.47/6.80 μM; IC50 ≥29 μM against PDE1/2/3/5/7/8/9/10/11 subtypes). Comparing to Rolipram, D159678 offers similar in vivo efficacy on long-term memory formation by novel object recognition test (MED = 3 μg/kg mouse iv., 1 μg/kg rat p.o.), while being more effective in the scopolamine-impaired Y-maze tests (D159678/Rolipram MED = 0.1/1 μg/kg mouse iv. or 30/100 μg/kg mouse p.o.) and much less emetic in shrews/dog/monkeys (by 100-/3000-/500-fold).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane S Sutcliffe et al.
PloS one, 9(7), e102449-e102449 (2014-07-23)
Cyclic adenosine monophosphate (cAMP) signalling plays an important role in synaptic plasticity and information processing in the hippocampal and basal ganglia systems. The augmentation of cAMP signalling through the selective inhibition of phosphodiesterases represents a viable strategy to treat disorders
David J Titus et al.
Neurobiology of learning and memory, 148, 38-49 (2018-01-03)
Traumatic brain injury (TBI) significantly decreases cyclic AMP (cAMP) signaling which produces long-term synaptic plasticity deficits and chronic learning and memory impairments. Phosphodiesterase 4 (PDE4) is a major family of cAMP hydrolyzing enzymes in the brain and of the four
Alex B Burgin et al.
Nature biotechnology, 28(1), 63-70 (2009-12-29)
Phosphodiesterase 4 (PDE4), the primary cAMP-hydrolyzing enzyme in cells, is a promising drug target for a wide range of conditions. Here we present seven co-crystal structures of PDE4 and bound inhibitors that show the regulatory domain closed across the active
Chong Zhang et al.
Scientific reports, 7, 40115-40115 (2017-01-06)
Inhibition of cyclic AMP (cAMP)-specific phosphodiesterase 4 (PDE4) has been proposed as a potential treatment for a series of neuropsychological conditions such as depression, anxiety and memory loss. However, the specific involvement of each of the PDE4 subtypes (PDE4A, 4B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service