Skip to Content
Merck

SML3192

GLX481304

≥98% (HPLC)

Synonym(s):

2-N-(3,4-Dimethylphenyl)-6-[[4-(3-methoxyphenyl)piperazin-1-yl]methyl]-1,3,5-triazine-2,4-diamine, N-(3,4-Dimethylphenyl)-6-[[4-(3-methoxyphenyl)-1-piperazinyl]methyl]-1,3,5-triazine-2,4-diamine, N2-(3,4-Dimethylphenyl)-6-[[4-(3-methoxyphenyl)-1-piperazinyl]methyl]-1,3,5-triazine-2,4-diamine, [4-Amino-6-[4-(3-methoxy-phenyl)-piperazin-1-ylmethyl]-1H-[1,3,5]triazin-2-ylidene]-(3,4-dimethyl-phenyl)-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H29N7O
CAS Number:
Molecular Weight:
419.52
NACRES:
NA.77
UNSPSC Code:
12352107
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

GLX481304 is a cell-permeable, noncytotoxic triazine-diamino compound that acts a potent, dual NOX2 and NOX4 isoform-selective inhibitor (IC50 ~ 1.25 μM) over NOX1. Does neither function as an antioxidant nor as a scavenger of reactive oxygen species (ROS). GLX481304 is shown to dose-dependently reduce ROS levels in hypoxia challenged mouse cardiomyocytes and improve contractile function in whole heart.
cell-permeable, noncytotoxic triazine-diamino compound that acts a potent, dual NOX2 and NOX4 isoform-selective inhibitor


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library