Skip to Content
Merck

SML3129

NIK SMI1

≥95% (HPLC)

Synonym(s):

(R)-6-(3-((3-Hydroxy-1-methyl-2-oxopyrrolidin-3-yl)-ethynyl)phenyl)-4-methoxypicolinamide, 6-[3-[2-[(3R)-3-Hydroxy-1-methyl-2-oxo-3-pyrrolidinyl]ethynyl]phenyl]-4-methoxy-2-pyridinecarboxamide, NIK small molecule inhibitor 1, NIK-SMI1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19N3O4
CAS Number:
Molecular Weight:
365.38
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(C1=NC(C2=CC(C#C[C@]3(C(N(CC3)C)=O)O)=CC=C2)=CC(OC)=C1)=O

InChI

1S/C20H19N3O4/c1-23-9-8-20(26,19(23)25)7-6-13-4-3-5-14(10-13)16-11-15(27-2)12-17(22-16)18(21)24/h3-5,10-12,26H,8-9H2,1-2H3,(H2,21,24)/t20-/m0/s1

InChI key

LQSHXYHWYGKAMX-FQEVSTJZSA-N

Biochem/physiol Actions

NIK SMI1 is an orally active, ATP site-targeting, potent and selective NF-κB-inducing kinase inhibitor (h/m NIK IC50 = 230/395 pM; hKHS1/hPKD1/hLRRK2 = 49.6/75.2/247.8 nM) that selectively disrupts noncanonical NF-κB signaling (anti-LTβR-induced p52 nuclear translocation IC50 = 70 nM; TNFα-induced p65 RelA nuclear translocation IC50 >2 μM). SMI1 inhibits BAFF receptor- and CD40-mediated singling in cultures (human and murine B-cells) and shows in vivo efficacy in the NZB/W F1 murine experimental lupus model (6-400 mg/kg/day, b.i.d. p.o.).
Orally active NF-κB-inducing kinase (NIK) inhibitor that selectively disrupts noncanonical NF-κB activation with in vivo efficacy in murine experimental lupus model.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library