Skip to Content
Merck
All Photos(1)

Key Documents

SML3102

Sigma-Aldrich

Ceftaroline fosamil acetate hydrate

≥98% (HPLC)

Synonym(s):

4-(2-((6R,7R)-2-Carboxy-7-((Z)-2-(ethoxyimino)-2-(5-(phosphonoamino)-1,2,4-thiadiazol-3-yl)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-ylthio)thiazol-4-yl)-1-methylpyridinium acetate, hydrate, Ceftaroline fosamil acetate hydrate, PPI 0903 hydrate, PPI-0903 hydrate, PPI0903 hydrate, TAK 599 hydrate, TAK-599 hydrate, TAK599 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N8O8PS4 · C2H3O2 · xH2O
CAS Number:
Molecular Weight:
744.74 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

C[N+]1=CC=C(C=C1)C2=CSC(SC(CS[C@@]3([C@@H]4NC(/C(C5=NSC(NP(O)(O)=O)=N5)=N\OCC)=O)[H])=C(N3C4=O)C(O)=O)=N2.CC([O-])=O

InChI

1S/C22H21N8O8PS4.C2H4O2/c1-3-38-26-13(16-25-21(43-28-16)27-39(35,36)37)17(31)24-14-18(32)30-15(20(33)34)12(9-40-19(14)30)42-22-23-11(8-41-22)10-4-6-29(2)7-5-10;1-2(3)4/h4-8,14,19H,3,9H2,1-2H3,(H4-,24,25,27,28,31,33,34,35,36,37);1H3,(H,3,4)/b26-13-;/t14-,1

InChI key

UGHHNQFYEVOFIV-VRDMTWHKSA-N

Biochem/physiol Actions

Ceftaroline fosamil (PPI-0903, TAK-599) is a N-phosphono type prodrug of ceftaroline (cephalosporin 2a, PPI-0903M, T-91825), a fifth-generation penicillin-binding protein (PBP)-targeting cephalosporin class beta-lactam (β-lactam) antibiotic with broad-spectrum activity against gram-positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA) and extensively-resistant strains, such as vancomycin-intermediate S. aureus (VISA), heteroresistant VISA (hVISA), and vancomycin-resistant S. aureus (VRSA).
N-phosphono type prodrug of ceftaroline, a fifth-generation PBP-targeting cephalosporin antibiotic against gram-positive bacteria, including MRSA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Andes et al.
Antimicrobial agents and chemotherapy, 50(4), 1376-1383 (2006-03-30)
PPI-0903 is a new cephalosporin with broad-spectrum activity, including beta-lactam-resistant Streptococcus pneumoniae and Staphylococcus aureus. We used the neutropenic murine thigh and lung infection models to examine the pharmacodynamic characteristics of PPI-0903. Serum drug levels following four fourfold-escalating single doses
Evaluation of the in vivo efficacy of intramuscularly administered ceftaroline fosamil, a novel cephalosporin, against a methicillin-resistant Staphylococcus aureus strain in a rabbit endocarditis model.
Cédric Jacqueline et al.
The Journal of antimicrobial chemotherapy, 65(10), 2264-2265 (2010-08-19)
Tomoyasu Ishikawa et al.
Bioorganic & medicinal chemistry, 11(11), 2427-2437 (2003-05-09)
Crystalline 1 (TAK-599) is a novel N-phosphono prodrug of anti-methicillin-resistant Staphylococcus aureus (MRSA) cephalosporin 2a (T-91825) that has high affinity for penicillin-binding protein (PBP) 2' (IC(50); 0.90 microg/mL) and shows potent in vitro anti-MRSA activity (MIC against MRSA N133; 1.56
Christopher Duplessis et al.
Clinical medicine reviews in therapeutics, 3 (2011-07-26)
Microbial resistance has reached alarming levels, threatening to outpace the ability to counter with more potent antimicrobial agents. In particular, methicillin-resistant Staphylococcus aureus (MRSA) has become a leading cause of skin and soft-tissue infections and PVL-positive strains have been associated
Cédric Jacqueline et al.
Antimicrobial agents and chemotherapy, 51(9), 3397-3400 (2007-06-27)
Using the rabbit endocarditis model, we compared the activity of a new broad-spectrum cephalosporin, ceftaroline, with those of linezolid and vancomycin against methicillin-resistant Staphylococcus aureus. After a 4-day treatment, ceftaroline exhibited superior bactericidal in vivo activity against resistant S. aureus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service