Skip to Content

SML2991

Ru265

≥95% (HPLC), powder, mitochondrial calcium uniporter inhibitor

Synonym(s):

Ru2(μ-N)(NH3)8Cl2]Cl3, trans,trans-Octaamminedichloro-μ-nitridodi-Ruthenium(3+) trichloride, Nitride, ruthenium complex

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Cl5H24N9Ru2
CAS Number:
Molecular Weight:
529.66
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ru265, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to dark beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl[Ru](N)(N)(N)(N)n[Ru](N)(N)(N)(N)Cl

Biochem/physiol Actions

Ru265 is a non-cytotoxic, aqueous soluble nitrido-bridged dinuclear Ru(IV) complex that acts as a selective mitochondrial calcium uniporter (MCU) inhibitor via a manner distinct from the oxo-bridged Ru360. Ru265 selectively reduces 100 μM histamine-induced mitochondrial [Ca2+] (mCa2+), but not cytosolic [Ca2+] (cCa2+) transient (50 ?M; HeLa) by blocking MCU-mediated mCa2+ uptake at a >10-fold greater potency as Ru360 (IC50 ~2.6 nM vs. 30 nM; HEK293T) without effecting mCa2+ efflux or membrane potential (ΔΨm). Ru265 (50 μM) protects neonatal rat ventricular myocytes from hypoxia/reoxygenation injury by preventing mPTP opening and mCa2+ overload.
Selective mitochondrial calcium uniporter (MCU) inhibitor that blocks mitochondrial Ca2+ uptake at a >10-fold greater potency as Ru360.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library