Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C30H39N6O9P
CAS Number:
Molecular Weight:
658.64
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
[P](=O)(N[C@@H](C)C(=O)OCC(C)(C)C)(OC[C@H]3O[C@H]([C@@]([C@@H]3O)(O)C)[n]4c5[nH][c](nc(c5nc4)OC)=N)Oc1c2c(ccc1)cccc2
InChI
1S/C30H39N6O9P/c1-17(26(38)42-15-29(2,3)4)35-46(40,45-20-13-9-11-18-10-7-8-12-19(18)20)43-14-21-23(37)30(5,39)27(44-21)36-16-32-22-24(36)33-28(31)34-25(22)41-6/h7-13,16-17,21,23,27,37,39H,14-15H2,1-6H3,(H,35,40)(H2,31,33,34)/t17-,21+,23+,27+,30+,46?/m0/s1
InChI key
YFXGICNMLCGLHJ-RSKRLRQZSA-N
Biochem/physiol Actions
BMS-986094 is an orally available prodrug of nucleotide analogue 2′-C-methylguanosine that potently inhibits Hepatitis C virus (HCV) replication. It is a guanosine nucleotide inhibitor of HCV RNA-dependent RNA polymerase NS5b.
guanosine nucleotide inhibitor of HCV RNA-dependent RNA polymerase NS5b
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.