Skip to Content
Merck

Skip To

SML2873

EIDD-2801

≥98% (HPLC), ribonucleoside analog, powder

Synonym(s):

((2R,3S,4R,5R)-3,4-Dihydroxy-5-(4-(hydroxyamino)-2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl isobutyrate, (oh)4Cyd lester, 4-N-HO-Cyd ester, BHC ester, EIDD 1931 5′-isopropylester, EIDD 1931 prodrug, EIDD 2801, EIDD-1931 5′-isopropylester, EIDD-1931 prodrug, EIDD1931 5′-isopropylester, EIDD1931 prodrug, EIDD2801, N4-Hydroxycytidine 5′-isopropylester, N4-OHcyd ester, N4OHcyd ester, NHC ester, beta-D-N4-Hydroxycytidine 5′-isopropylester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H19N3O7
CAS Number:
Molecular Weight:
329.31
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EIDD-2801, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

Broad antiviral N4-hydroxycytidine (EIDD-1931) prodrug with in vivo efficacy and enhanced oral bioavailability.
EIDD-2801 is an orally active, 5′-isopropylester prodrug form of the antiviral ribonucleoside analog N4-hydroxycytidine (NHC, EIDD-1931) with known anti-viral activity against SARS-CoV, SARS-CoV-2, RSV, influenza A & B (IAV & IBV), HCV, pestivirus, bovine viral diarrhoea virus (BVDV), Ebola (EBOV), Chikungunya (CHIKV), venezuelan equine encephalitis (VEEV). EIDD-2801 is efficiently hydrolyzed to NHC in vivo, being more orally bioavailable than NHC in nonhuman primates and ferrets, while maintaining similar oral bioavailability as NHC in mice. EIDD-2801 displays in vivo efficacy against pandemic and seasonal IAV strains in ferrets (lowest ED = 2.3 and 7 mg/kg b.i.d. p.o., respectively).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2872SML0688A2846
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC), solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 1 mg/mL, clear (warmed)

solubility

H2O: soluble

color

white to beige

color

white to beige

color

faintly pink to red

color

off-white to light tan


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters