Skip to Content
Merck
All Photos(1)

Key Documents

SML2804

Sigma-Aldrich

Dapagliflozin

≥98% (HPLC), powder, SGLT2 inhibitor

Synonym(s):

(1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol, (2S,3R,4R,5S,6R)-2-(3-(4-Ethoxybenzyl)-4-chlorophenyl)-6-hydroxymethyl-tetrahydro-2H-pyran-3,4,5-triol, BMS 512148, BMS-512148

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClO6
CAS Number:
Molecular Weight:
408.87
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

product name

Dapagliflozin, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D (+7.0° to +13.0°, c = 0.2 in methanol)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCOC1=CC=C(C=C1)CC2=C(C=CC([C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)CO)O)O)O)=C2)Cl

InChI

1S/C21H25ClO6/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21+/m1/s1

InChI key

JVHXJTBJCFBINQ-ADAARDCZSA-N

Biochem/physiol Actions

Dapagliflozin (BMS-512148) is an orally active, potent and selective renal sodium-dependent glucose cotransporter 2 (SGLT2; SLC5A2) inhibitor (human/rat SGLT2 IC50 = 1.1/3 nM vs. human/rat SGLT1 IC50 = 1.39/0.6 μM; R-methyl-D-glucopyranoside (AMG) uptake assay with respective CHO transfectants). Dapagliflozin reduces blood glucose levels (by 55% in streptozotocin (STZ)-induced hyperglycemic rats 5 hrs post 0.1 mg/kg p.o.) by preventing the renal glucose reabsorption process and promoting glucose excretion in urine with good pharmacokinetic properties and oral availability (84% in rats and 83% in dogs, respectively, post 1 mg/kg and 6.6 mg/kg p.o.) in vivo.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - STOT RE 1

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Yoshioka et al.
Obesity research & clinical practice, 13(5), 505-510 (2019-08-31)
Chronopharmacology is the study of the varying responses of drugs to changes in biological timing and endogenous periodicities. The selective sodium-glucose cotransporter 2 inhibitor, dapagliflozin, is a globally prescribed antihyperglycemic drug. Although dapagliflozin is usually administered once a day, the
Kazuno Omori et al.
Metabolism: clinical and experimental, 98, 27-36 (2019-06-17)
To explore the beneficial effects of dapagliflozin and/or insulin glargine on the pancreatic beta cell mass and hepatic steatosis in db/db mice. Six-week-old db/db mice were assigned to one of four groups: untreated (Placebo), treated with dapagliflozin (Dapa), treated with
Ryo Shibusawa et al.
Scientific reports, 9(1), 9887-9887 (2019-07-10)
The new type 2 diabetes drug, dapagliflozin, reduces blood glucose levels and body weight by inhibiting sodium glucose transporter 2 (SGLT2) in proximal tubular cells. SGLT2 inhibitors might modulate glucose influx into renal tubular cells, thereby regulating the metabolic conditions
Fengjuan Huang et al.
Frontiers in endocrinology, 10, 441-441 (2019-07-25)
Tubulointerstitial fibrosis (TIF) plays an important role in the progression of renal fibrosis in diabetic nephropathy (DN). Accumulating evidence supports a crucial inhibitory effect of dapagliflozin, a SGLT2 inhibitor, on TIF, but the underlying mechanisms remain largely unknown. This study
Nannan Zhang et al.
Cardiovascular diabetology, 18(1), 107-107 (2019-08-21)
Heart failure with preserved ejection fraction (HFpEF) is a difficult disease with high morbidity and mortality rates and lacks an effective treatment. Here, we report the therapeutic effect of dapagliflozin, a sodium-glucose cotransporter 2 inhibitor (SGLT2i), on hypertension + hyperlipidemia-induced HFpEF in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service