Skip to Content
Merck
All Photos(1)

Documents

SML2499

Sigma-Aldrich

TY-52156

≥98% (HPLC)

Synonym(s):

1-(4-Chlorophenylamino)-1-(4-chlorophenylhydrazono)-3,3-dimethyl-2-butanone, 1-(4-Chlorophenylhydrazono)-1-(4-chlorophenylamino)-3,3-dimethyl-2-butanone, N-(4-Chloroanilino)-N′-(4-chlorophenyl)-3,3-dimethyl-2-oxobutanimidamide, TY 52156, TY52156

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19Cl2N3O
CAS Number:
Molecular Weight:
364.27
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)(C(/C(NC1=CC=C(C=C1)Cl)=N/NC2=CC=C(C=C2)Cl)=O)C

InChI

1S/C18H19Cl2N3O/c1-18(2,3)16(24)17(21-14-8-4-12(19)5-9-14)23-22-15-10-6-13(20)7-11-15/h4-11,22H,1-3H3,(H,21,23)

InChI key

XONRRGIRSGNWFP-UHFFFAOYSA-N

Biochem/physiol Actions

TY-52156 is a potent sphingosine 1-phosphate (S1P) receptor 3 (S1P3; S1PR3) antagonist that selectively inhibits S1P-stimulated Ca2+ response in human S1P3-expressing cells (Ki = 110 nM) and Eu-GTP binding to S1P3-containing membranes over those of S1P1/2/4/5. TY-52156 inhibits S1P3-dependent signaling events in human coronary artery smooth muscle cells (1 μM), S1P3-dependent coronary flow decrease in perfused rat hearts, as well as S1P3-mediated acute mechanical pain in mice (10 nmol/20 μL/paw id.) and bradycardia in anesthetized rats (30 mg/kg TY-52156 p.o.) in vivo with good oral bioavailability (F = 70.9%; 1 mg/kg p.o. in rats).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Murakami et al.
Molecular pharmacology, 77(4), 704-713 (2010-01-26)
Sphingosine 1-phosphate (S1P) induces diverse biological responses in various tissues by activating specific G protein-coupled receptors (S1P(1)-S1P(5) receptors). The biological signaling regulated by S1P(3) receptor has not been fully elucidated because of the lack of an S1P(3) receptor-specific antagonist or
Shushu Wang et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 1912-1923 (2018-04-02)
BACKGROUND Triple negative breast cancer (TNBC) has a more aggressive recurrence. Previous reports have demonstrated that sphingosine kinase 1 (SphK1) is a crucial regulator of breast cancer progression. However, the correlation of SphK1 with clinical prognosis has been poorly investigated.
Iuliia Filipenko et al.
Biochimica et biophysica acta, 1861(11), 1840-1851 (2016-09-13)
Breast cancer is one of the most common and devastating malignancies among women worldwide. Recent evidence suggests that malignant progression is also driven by processes involving the sphingolipid molecule sphingosine 1-phosphate (S1P) and its binding to cognate receptor subtypes on
Xiao-Jing Sun et al.
Journal of cellular and molecular medicine, 22(3), 1769-1777 (2017-11-24)
Cumulating evidences suggested an important role of sphingosine-1-phosphate (S1P) and its receptors in regulating endothelial barrier integrity. Our previous study revealed that the circulating S1P levels and renal expression of S1PRs correlated with disease activity and renal damage in patients
Zhijing Zhao et al.
Experimental and therapeutic medicine, 15(6), 5007-5016 (2018-05-29)
Thyroid carcinoma is characterized by an aggressive behavior, lack of effective targeted therapies and a high rate of relapse. Sphingosine kinase 1 (SPHK1) has been reported to be a critical regulatory factor in the progression of thyroid carcinoma, but the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service