Skip to Content

SML2473

Pitavastatin calcium

≥98% (HPLC), HMG CoA reductase inhibitor, powder

Synonym(s):

(+)monocalcium bis{(3R, 5S, 6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolyl]-3,5dihydroxy-6-heptenoate}, (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic acid calcium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H23FNO4 · 0.5Ca
CAS Number:
Molecular Weight:
440.49
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pitavastatin calcium, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +19 to +24°, c = 1 in acetonitrile: water (1:1)

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Ca+2].Fc5ccc(cc5)c6c7c(nc(c6\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C8CC8)cccc7.Fc1ccc(cc1)c2c3c(nc(c2\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C4CC4)cccc3

InChI

1S/2C25H24FNO4.Ca/c2*26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31;/h2*1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31);/q;;+2/p-2/b2*12-11+;/t2*18-,19-;/m11./s1

InChI key

RHGYHLPFVJEAOC-FFNUKLMVSA-L

Biochem/physiol Actions

HMG CoA reductase inhibitor
Pitavastatin calcium is a water-soluble inhibitor of hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonic acid, the rate-limiting step in cholesterol biosynthesis. Pitavastatin calcium is antilipidemic and is used to reduce plasma cholesterol levels and prevent cardiovascular disease.


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library