Skip to Content
Merck
All Photos(1)

Key Documents

SML2452

Sigma-Aldrich

LY2444296

≥98% (HPLC)

Synonym(s):

(S)-3-Fluoro-4-(4-((2-(3-fluorophenyl)pyrrolidin-1-yl)methyl)phenoxy)benzamide, 3-Fluoro-4-[4-[[(2S)-2-(3-fluorophenyl)-1-pyrrolidinyl]methyl]phenoxy]benzamide, 4-[4-[[(2S)-2-(3-Fluorophenyl)-1-pyrrolidinyl]methyl]phenoxy]-3-fluorobenzamide, FP3FBZ, LY 2444296, LY-2444296

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22F2N2O2
CAS Number:
Molecular Weight:
408.44
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(C1=CC=C(OC2=CC=C(CN3CCC[C@H]3C4=CC=CC(F)=C4)C=C2)C(F)=C1)=O

InChI

1S/C24H22F2N2O2/c25-19-4-1-3-17(13-19)22-5-2-12-28(22)15-16-6-9-20(10-7-16)30-23-11-8-18(24(27)29)14-21(23)26/h1,3-4,6-11,13-14,22H,2,5,12,15H2,(H2,27,29)/t22-/m0/s1

InChI key

WHHMECUDUQRPJT-QFIPXVFZSA-N

Biochem/physiol Actions

Brain-penetrant, orally active, short-acting, high-affinity, potent and selective κ (kappa) opioid receptor (KOR) antagonist with in vitro and in vivo efficacy.
LY2444296 is a brain-penetrant, orally active, short-acting, high-affinity, potent and selective κ (kappa) opioid receptor (KOR) antagonist (human κ/μ/δ Ki = 0.565/35.8/211 nM against diprenorphine binding; GTP-γ-S binding IC50/agonist/subtype transfectant = 1.57 nM/300 nM U69593/κ, 21.3 nM/1 μM DAMGO/μ 293 nM/30 nM DPDPE/d). LY2444296 reverses κ agonist antinociceptive efficacy in vivo (ED50 = 0.24 mg/kg p.o. against 1 mg/kg U69593 sc. by rat formalin test), decreases immobility time (10 or 30 mg/kg sc.) and prevents enhanced alcohol consumption (5 mg/kg i.p.) among mice subjected to stress by forced swimming.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Valenza et al.
Psychopharmacology, 234(15), 2219-2231 (2017-05-28)
The recruitment of the stress circuitry contributes to a shift from positive to negative reinforcement mechanisms sustaining long-term cocaine addiction. The kappa opioid receptor (KOPr) signaling is upregulated by stress and chronic cocaine exposure. While KOPr agonists induce anhedonia and
Rachel I Anderson et al.
Frontiers in cellular neuroscience, 10, 45-45 (2016-03-05)
Our laboratory has previously demonstrated that daily forced swim stress (FSS) prior to ethanol drinking sessions facilitates enhanced ethanol consumption in mice with a history of chronic intermittent ethanol (CIE) vapor exposure without altering ethanol intake in air-exposed controls. Because
Brian Reed et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 43(4), 739-750 (2017-09-01)
The κ-opioid receptor (KOP-r) system and its endogenous ligands, the dynorphins, are involved in the neurobiological regulation of addictive states, and of mood. There are limited data on the impact of selective KOP-r antagonism in humans on basic biobehavioral functions
Charles H Mitch et al.
Journal of medicinal chemistry, 54(23), 8000-8012 (2011-10-01)
Arylphenylpyrrolidinylmethylphenoxybenzamides were found to have high affinity and selectivity for κ opioid receptors. On the basis of receptor binding assays in Chinese hamster ovary (CHO) cells expressing cloned human opioid receptors, (S)-3-fluoro-4-(4-((2-(3-fluorophenyl)pyrrolidin-1-yl)methyl)phenoxy)benzamide (25) had a K(i) = 0.565 nM for
Peng Huang et al.
Neuroscience letters, 615, 15-20 (2016-01-19)
Prototypical long-acting kappa opioid receptor (KOPR) antagonists [e.g., norbinaltorphimine (norBNI)] have been reported to exert anxiolytic-like effects in several commonly used anxiety tests in rodents including the novelty-induced hypophagia (NIH) and elevated plus maze (EPM) tests. It remains unknown if

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service